Skip to Content
Merck
All Photos(1)

Key Documents

NA61

Sigma-Aldrich

Anti-BrdU (Ab-3) Mouse mAb (Mobu-1)

liquid, clone Mobu-1, Calbiochem®

Synonym(s):

Anti-Bromodeoxyuridine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Mobu-1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-BrdU (Ab-3), mouse monoclonal, clone Mobu-1, recognizes BrdU. Requires biosynthetic labeling of the target cells with BrdU. It is validated for ICC and use with frozen and paraffin sections.
Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with P3-X63-Ag8-653 mouse myeloma cells (see application references). Recognizes BrdU (bromodeoxyuridine).
Recognizes BrdU (bromodeoxyuridine). Use of this antibody first requires biosynthetic labeling of the target cells with bromodeoxyuridine. This is the preferred BrdU antibody for staining.

Immunogen

BrdU-labeled DNA

Application


Frozen Sections (2 g/ml)
Immunocytochemistry (1 g/ml)
Paraffin Sections (2.5 g/ml, trysin pre-treatment required)
Immunoblotting (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.

Analysis Note

Negative Control
Unlabeled cells
Positive Control
BrdU labeled DNA

Other Notes

Gratzner, H.G. 1982. Science218, 474.
Gratzner, H.G. and Leif, R.C. 1981. Cytometry1, 385.
Gratzner, H.G. et al. 1975. Exp. Cell Res.95, 88.
To enable antibody binding to the incorporated BrdU, cells must be fixed, permeabilized, and the DNA denatured. This is done in one step by treatment with Fixative/Denaturing Solution (70% EtOH, 0.1 N NaOH). Use of this antibody first requires biosynthetic labeling of the target cells with bromodeoxyuridine (Cat. No. 203806). Cultured cells can be pulse labeled with 10 µM bromodeoxyuridine for 30 minutes. For whole tissues, animals can be injected with 50 mg BrdU/kg body weight, sacrificed 1 h later, and the target organ(s) removed. Antibody should be titrated for optimal results in individual system.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick E Nyman et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(24), 6308-6318 (2018-08-09)
Head and neck squamous cell carcinoma (HNSCC), a common cancer worldwide, is etiologically associated with tobacco use, high alcohol consumption, and high-risk human papillomaviruses (HPV). The Notch signaling pathway, which is involved in cell differentiation decisions with differential downstream targets
Shali Chen et al.
Molecular medicine reports, 22(1), 353-361 (2020-04-23)
20(S)‑Protopanaxadiol (PPD) is an active ginseng metabolite and is the final form of protopanaxadiol saponins metabolized by human intestinal microflora. The neuroprotective effects and mechanisms underlying PPD on neural stem cells (NSCs) are not completely understood. The aim of the
Mieke Baan et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 309(7), R788-R794 (2015-07-24)
Transgenic mouse models are designed to study the role of specific proteins. To increase transgene expression the human growth hormone (hGH) minigene, including introns, has been included in many transgenic constructs. Until recently, it was thought that the hGH gene
Ian B DeMeritt et al.
Virology, 346(1), 15-31 (2005-11-24)
Infection of fibroblasts by human cytomegalovirus (HCMV) rapidly activates the NF-kappaB signaling pathway, which we documented promotes efficient transactivation of the major immediate-early promoter (DeMeritt, I.B., Milford, L.E., Yurochko, A.D. (2004). Activation of the NF-kappaB pathway in human cytomegalovirus-infected cells
Sathish Kumar Mungamuri et al.
Cancer research, 66(9), 4715-4724 (2006-05-03)
Notch signaling is believed to promote cell survival in general. However, the mechanism is not clearly understood. Here, we show that cells expressing intracellular domain of human Notch1 (NIC-1) are chemoresistant in a wild-type p53-dependent manner. NIC-1 inhibited p53 by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service