Skip to Content
Merck
All Photos(3)

Key Documents

E2764

Sigma-Aldrich

Anti-Endothelin Receptor B antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ET-B, Anti-ET-Specific Endothelin Receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100
western blot (chemiluminescent): 1:200

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Endothelin Receptor B (EDNRB) is a G-protein coupled receptor and that consists of seven transmembrane domains. EDNRB gene is located on the human chromosome at 13q22.3.

Immunogen

synthetic peptide corresponding to amino acids 298-313 of rat endothelin receptor type B. The epitope is identical in all known vertebrate ET-B receptors

Application

Anti-Endothelin Receptor B antibody produced in rabbit has been used in immunohistochemistry (1:100).

Biochem/physiol Actions

EDNRB Endothelin receptor type B (ETB) plays a pivotal role in oesophageal cancer and may act as a therapeutic target for this intractable malignancy. It has a novel role in oligodendroglioma proliferation and acts as a potential therapeutic alternative for oligodendrogliomas. Lower expression of ETB receptor responsiveness is associated with polycystic ovary syndrome (PCOS) and may reflect lower endothelial-mediated vasodilatation independent of generally lower vascular reactivity.
Mutations in the EDNRB gene are associated with Hirschsprung disease.

Physical form

Lyophilized at 0.8 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Amiel et al.
Human molecular genetics, 5(3), 355-357 (1996-03-01)
Hirschsprung disease (HSCR, aganglionic megacolon) is a frequent congenital malformation regarded as a multigenic neurocristopathy. Two susceptibility genes have been recently identified in HSCR, namely the RET proto-oncogene and the endothelin B receptor (EDNRB) gene. Hitherto however, homozygosity for EDNRB
M K Shin et al.
Nature, 402(6761), 496-501 (1999-12-11)
Endothelin receptor B (EDNRB) is a G-protein-coupled receptor with seven transmembrane domains which is required for the development of melanocytes and enteric neurons. Mice that are homozygous for a null mutation in the Ednrb gene are almost completely white and
Chun-Yun Wu et al.
Glia, 54(6), 513-525 (2006-08-10)
Amoeboid microglial cells (AMC) which transiently exist in the corpus callosum in the postnatal rat brain expressed endothelins (ETs), specifically endothelin-1 (ET-1) and ET3 as revealed by real time RT-PCR. ET immunoreactive AMC occurred in large numbers at birth, but
Victor M Salinas-Torres et al.
Pediatric surgery international, 34(9), 931-943 (2018-08-11)
Gastroschisis has been assumed to have a low rate of syndromic and primary malformations. We aimed to systematically review and explore the frequency and type of malformations/chromosomal syndromes and to identify significant biological/genetic roles in gastroschisis. Population-based, gastroschisis-associated anomalies/chromosomal defects
T Sakurai et al.
Nature, 348(6303), 732-735 (1990-12-20)
Endothelin-1 was initially identified as a 21-residue potent vasoconstrictor peptide produced by vascular endothelial cells, but was subsequently found to have many effects on both vascular and non-vascular tissues. The discovery of three isopeptides of the endothelin family, ET-1, ET-2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service