Skip to Content
Merck
All Photos(1)

Documents

A9378

Sigma-Aldrich

L-Amino Acid Oxidase from Crotalus adamanteus

Type IV, ≥4.0 units/mg protein, aqueous suspension

Synonym(s):

L-AAO, L-Amino acid:oxygen oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type IV

Quality Level

form

aqueous suspension

specific activity

≥4.0 units/mg protein

mol wt

~130 kDa

contains

toluene as preservative

concentration

≥5.0 mg/mL

solubility

H2O: soluble 1.0 mg/mL, clear

storage temp.

2-8°C

Application

L-amino acid oxidase (LAAO) is used to convert L-amino acids to their corresponding α-keto acids. L-amino acid oxidase, from Sigma, has been used in leucine aminopeptidase (LAP) activity assays. 35S dimethylsulfoniopropionate (DMSP) has been synthesized chemically from 35S L-methionine using LAAO from Sigma to form 35S 3-methiolpropionate.

Biochem/physiol Actions

L-Amino acid oxidase is a flavoprotein with a molecular weight of 130 kDa. It consists of two different subunits of approximately 70,000 Da. Each molecule of holoenzyme has two FAD molecules. It is a glycoprotein containing about 2-5% carbohydrate, including sialic acid. Optimum pH is approximately 7.5.The enzyme may be reversibly inactivated by incubation in phosphate buffer, pH 7.5 at 38 °C. L-amino acid oxidase is involved in various metabolic pathways such as alanine and aspartate metabolism, methionine metabolism, valine, leucine and isoleucine degradation, tyrosine metabolism, phenylalanine metabolism, tryptophan metabolism, phenylalanine, tyrosine and tryptophan biosynthesis, and alkaloid biosynthesis. It occurs in many snake venoms apart from microorganisms and animal tissue, especially in kidney and liver.

Packaging

Package size based on protein content

Unit Definition

One Unit oxidizes one micromole of L-leucine per minute at 25 °C, pH 7.6

Preparation Note

Dissolves in water at 1 mg/mL concentration to form a clear solution.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and enumeration of bacteria assimilating dimethylsulfoniopropionate (DMSP) in the North Atlantic and Gulf of Mexico
Malmstrom RR, et al.
Limnology and Oceanography, 49(2), 597-606 (2004)
Stefania Digiovanni et al.
Scientific reports, 10(1), 10135-10135 (2020-06-25)
Reactive Intermediate Deaminase (Rid) protein superfamily includes eight families among which the RidA is conserved in all domains of life. RidA proteins accelerate the deamination of the reactive 2-aminoacrylate (2AA), an enamine produced by some pyridoxal phosphate (PLP)-dependent enzymes. 2AA
Jiro Arima et al.
The Journal of biological chemistry, 281(9), 5885-5894 (2006-01-13)
Streptomyces griseus leucine aminopeptidase (SGAP), which has two zinc atoms in its active site, is clinically important as a model for understanding the structure and mechanism of action of other metallopeptidases. SGAP is a calcium-activated and calcium-stabilized enzyme, and its
Wen Ming Chen et al.
Enzyme and microbial technology, 49(4), 372-379 (2011-11-25)
A brownish yellow pigmented bacterial strain, designated antisso-27, was recently isolated from a water area of saltpan in Southern Taiwan. Phylogenetic analyses based on 16S rRNA gene sequences indicate that strain antisso-27 belongs the genus Aquimarina in the family Flavobacteriacea
Jaro T Nuutinen et al.
Microbiology (Reading, England), 158(Pt 1), 272-283 (2011-10-15)
Catabolism of amino acids is a central process in cellular nitrogen turnover, but only a few of the mechanisms involved have been described from basidiomycete fungi. This study identified one such mechanism, the l-amino acid oxidase (Lao1) enzyme of Hebeloma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service