Skip to Content
Merck
All Photos(2)

Documents

A7419

Sigma-Aldrich

Avidin–Peroxidase

buffered aqueous solution

Synonym(s):

Avidin–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

avidin from egg white
enzyme from horseradish

Quality Level

conjugate

peroxidase conjugate

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:100,000
western blot: 1:400,000-1:800,000 using detecting β-Actin in total cell extract of HeLa cells (5-10 μg/well)

shipped in

dry ice

storage temp.

−20°C

General description

Avidin is homotetrameric protein (68kDa) obtained from egg whites and binds strongly to biotin. Thus, avidin-biotin association has been utilized in immunoassays to detect the localization of antigens in tissues. The use of avidin-biotin immunoassay enhances the sensitivity of the technique and facilitates the detection of antigens in low quantities.

Application

Avidin-Peroxidase has been used for ELISA applications. The product can also be used for western blot applications at dilutions ranging from 1:400,000 to 1:800,000.
Western blot analysis of proteins in HUVEC cells that had underwent cell surface biotinylation were detected using HRP-conjugated avidin at 33.3 ng/ml.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT.

Preparation Note

Affinity purified protein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clare Baecher-Allan et al.
Journal of immunology (Baltimore, Md. : 1950), 169(11), 6210-6217 (2002-11-22)
CD4(+)CD25(+high) T cells are potent regulators of autoreactive T cells. However, it is unclear how regulatory CD4(+)CD25(+high) cells discriminate between desirable inflammatory immune responses to microbial Ags and potentially pathologic responses by autoreactive T cells. In this study, an in
Prajna Lalitha et al.
American journal of ophthalmology, 144(4), 552-556 (2007-08-19)
To study the range of ocular symptoms in a cohort of patients with chikungunya infection. Retrospective, observational case series. Patients attending a tertiary eye care hospital in South India were included in the study. We included adult patients with serologically
Nir Yogev et al.
Cell reports, 38(13), 110565-110565 (2022-03-31)
Interleukin (IL)-10 is considered a prototypical anti-inflammatory cytokine, significantly contributing to the maintenance and reestablishment of immune homeostasis. Accordingly, it has been shown in the intestine that IL-10 produced by Tregs can act on effector T cells, thereby limiting inflammation. Herein
J J Quinlan et al.
Applied and environmental microbiology, 63(2), 482-487 (1997-02-01)
Five monoclonal antibodies against bacterial spores of Bacillus cereus T and Clostridium sporogenes PA3679 were developed. Two antibodies (B48 and B183) were selected for their reactivity with B. cereus T spores, two (C33 and C225) were selected for their reactivity
Dominika Lukas et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(8), E1480-E1489 (2017-02-09)
TGF-β is an anti-inflammatory cytokine whose signaling is negatively controlled by Smad7. Previously, we established a role for Smad7 in the generation of autoreactive T cells; however, the function of Smad7 in dendritic cells (DCs) remains elusive. Here, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service