Skip to Content
Merck
All Photos(5)

Key Documents

H8125

Sigma-Aldrich

L-Histidine monohydrochloride monohydrate

≥98% (HPLC)

Synonym(s):

L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98% (HPLC)

mp

254 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL, clear, colorless to very faintly yellow

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Histidine monohydrochloride monohydrate has been used as a component of broth to match the tuna meat amino acid composition. It has also been used for partial rescue of the IMPL2 (myo-inositol monophosphatase) phenotype during seed development of Arabidopsis.

Biochem/physiol Actions

Histidine takes part in protein methylation. It is associated with the hemoglobin structure and function. Histidine is present in antioxidative dipeptides. It is involved in one-carbon unit metabolism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Sato et al.
Journal of plant research, 124(3), 385-394 (2010-10-21)
Myo-inositol monophosphatase (IMP) catalyzes the dephosphorylation of myo-inositol 3-phosphate in the last step of myo-inositol biosynthesis. IMP is also important in phosphate metabolism and is required for the biosynthesis of cell wall polysaccharides, phytic acid, and phosphatidylinositol. In Arabidopsis, IMP
Expression and functions of myo-inositol monophosphatase family genes in seed development of Arabidopsis
Sato Y, et al.
Journal of Plant Research, 124(3), 385-394 (2011)
Modelling the effect of temperature, carbon dioxide, water activity and pH on growth and histamine formation by Morganella psychrotolerans
Emborg J and Dalgaard P
International Journal of Food Microbiology, 128(2), 226-233 (2008)
Garrett RH and Grisham CM
Biochemistry (2012)
Kiana Khadem-Abbassi et al.
Nanomaterials (Basel, Switzerland), 9(5) (2019-05-07)
This work describes the preparation of molecularly imprinted polymer (MIP)-modified core/shell CdTe0.5S0.5/ZnS quantum dots (QDs). The QDs@MIP particles were used for the selective and sensitive detection of dopamine (DA). Acrylamide, which is able to form hydrogen bonds with DA, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service