A6237
Apyrase from potato
recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Synonym(s):
Apyrase from potato, Adenosine 5′-diphosphatase, Adenosine 5′-triphosphatase
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Recommended Products
recombinant
expressed in Pichia pastoris
Quality Level
form
lyophilized powder
ATPase activity
≥1000 units/mg protein
shipped in
wet ice
storage temp.
−20°C
General description
Apyrase, also known as ATP-diphosphohydrolase, is expressed in wide variety of plant and animal tissues. The commercially accessible apyrase enzyme is isolated from potato tubers.
Biochem/physiol Actions
Apyrase hydrolyses the phosphoanhydride bonds of nucleoside tri- and di-phosphates in the presence of divalent cations. It has wide substrate specificity for nucleotides. This property of the enzyme makes it suitable for different biotechnical applications, including DNA sequencing and platelet-aggregation inhibition.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Subin Mao et al.
Lab on a chip, 21(16), 3128-3136 (2021-06-29)
Integrins are key players in platelet adhesion and aggregation. Integrin molecular tensions, the forces transmitted by integrin molecules, are regulated by both mechanical and biochemical cues, and the outside-in and inside-out signaling has been extensively studied. While the mechanical properties
Cloning, sequencing, and expression of a human brain ecto-apyrase related to both the ecto-ATPases and CD39 ecto-apyrases1.
Smith T M & Kirley T L
Biochimica et Biophysica Acta, 1386(1), 65-78 (1998)
Methylotrophic yeast Pichia pastoris as a host for production of ATP-diphosphohydrolase (apyrase) from potato tubers (Solanum tuberosum).
Nourizad N, et al.
Protein Expression and Purification, 27(2), 229-237 (2003)
Xuan Zhang et al.
European journal of medicinal chemistry, 199, 112397-112397 (2020-05-11)
Targeting BCL-XL via PROTACs is a promising strategy in reducing BCL-XL inhibition associated platelet toxicity. Recently, we reported potent BCL-XL PROTAC degraders that recruit VHL or CRBN E3 ligase. However, low protein expression or mutation of the responsible E3 ligase
Minfei Su et al.
Molecular cell, 80(1), 59-71 (2020-08-21)
Cardiac disease remains the leading cause of morbidity and mortality worldwide. The β1-adrenergic receptor (β1-AR) is a major regulator of cardiac functions and is downregulated in the majority of heart failure cases. A key physiological process is the activation of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service