Skip to Content
Merck
All Photos(2)

Key Documents

A5922

Sigma-Aldrich

Amifostine trihydrate

≥97% (TLC), powder

Synonym(s):

2-(3-Aminopropyl)aminoethyl phosphorothioate, WR2721

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H15N2O3PS · 3 H2O
CAS Number:
Molecular Weight:
268.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

OP(SCCNCCCN)(O)=O.O.O.O

InChI

1S/C5H15N2O3PS.3H2O/c6-2-1-3-7-4-5-12-11(8,9)10;;;/h7H,1-6H2,(H2,8,9,10);3*1H2

InChI key

TXQPXJKRNHJWAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amifostine may provide renal protection during PRRT using somatostatin analogs by mitigating radiation damage and reducing the absorbed kidney radiation dose. Amifostine remediates the degenerative effects of radiation on the mineralization capacity of the murine mandible.

Biochem/physiol Actions

Radioprotective agent. Selectively protects normal tissues from the damaging effects of anti-neoplastic radiation therapy. Selectivity is due to preferential uptake by normal tissues and subsequent metabolic activation to 2-(3-aminopropyl)aminoethanethiol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Capizzi et al.
Cancer, 72(11 Suppl), 3495-3501 (1993-12-01)
Amifostine (US Bioscience, West Conshohocken, PA; Ethyol, WR-2721), a phosphorylated thiol developed by the United States Army as a protective agent for military personnel in the event of nuclear warfare, has shown protection of normal tissues from the cytotoxic effects
David A Oʼdonovan et al.
Plastic and reconstructive surgery, 129(4), 636e-645e (2012-03-30)
The authors previously established an animal model of radiation-induced craniofacial bone growth inhibition and demonstrated the effectiveness of cytoprotection in preserving growth using amifostine, but the mechanism is unclear. The objective of this study was to investigate the acute and
Catherine N Tchanque-Fossuo et al.
Bone, 52(2), 712-717 (2012-08-14)
Adjuvant radiotherapy in the management of head and neck cancer remains severely debilitating. Fortunately, newly developed agents aimed at decreasing radiation-induced damage have shown great promise. Amifostine (AMF) is a compound, which confers radio-protection to the exposed normal tissues, such
Marleen Melis et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 53(5), 749-753 (2012-04-13)
Megalin-mediated renal retention of radiolabeled somatostatin analogs may lead to nephrotoxicity during peptide receptor radionuclide therapy (PRRT). The cytoprotective agent amifostine protected rats from long-term nephrotoxicity after PRRT with (177)Lu-DOTA,Tyr(3)-octreotate. This study describes the direct effect of amifostine on kidney
C M Spencer et al.
Drugs, 50(6), 1001-1031 (1995-12-01)
Amifostine (WR-2721) was originally developed as a radioprotective agent. In animals, it protects normal tissues from the damaging effects of irradiation and, as shown in more recent studies, of several cytotoxic agents. Protection of tumours is generally reduced compared with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service