Skip to Content
Merck
All Photos(1)

Key Documents

W2020

Sigma-Aldrich

WR-1065

≥98% (HPLC)

Synonym(s):

2-[(3-Aminopropyl)amino]ethanethiol dihydrochloride, WR-1065 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H14N2S · 2HCl
CAS Number:
Molecular Weight:
207.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

H2O: ≥20 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.NCCCNCCS

InChI

1S/C5H14N2S.2ClH/c6-2-1-3-7-4-5-8;;/h7-8H,1-6H2;2*1H

InChI key

XDRLRDHLCIFZIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

WR-1065 is cytoprotective cell-permeable reactive oxygen species scavenger and p53 activator and re-activator. Recently shown to have antiretroviral activity and an active metabolite of Amifostine which selectively protects normal tissues from the damaging effects of anti-neoplastic radiation therapy.
WR-1065 is the active metabolite of the prodrug amifostine, generated by alkaline phosphatase. It is found to accumulate in many epithelial tissues. WR-1065 protects cellular membranes and DNA from free radical induced damage.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J S Murley et al.
Free radical biology & medicine, 51(10), 1918-1925 (2011-09-29)
Manganese superoxide dismutase (SOD2)-mediated adaptive processes that protect against radiation-induced micronucleus formation can be induced in cells after a 2-Gy exposure by previously exposing them to either low-dose ionizing radiation (10cGy) or WR1065 (40μM), the active thiol form of amifostine.
Zaki S Badawy et al.
Fertility and sterility, 91(1), 173-178 (2008-01-22)
To determine the effect of 4-hydroperoxycyclophosphamide (4OOH-CP) on the respiration of human sperm, and investigate the protective properties of mesna and WR-1065. SUNY Upstate Medical University, Syracuse, NY. Men (n = 12) visited the Andrology Department for fertility evaluation. None.
Sarala Pamujula et al.
International journal of radiation biology, 84(11), 900-908 (2008-11-19)
N-(2-mercaptoethyl)1,3-diaminopropane (WR-1065), is the active metabolite of amifostine, a broad spectrum cytoprotective agent used in conjunction with both chemo- and radiotherapy of certain cancers. This report describes for the first time an oral formulation of WR-1065 and follows on from
David R Cassatt et al.
International journal of radiation oncology, biology, physics, 61(3), 901-907 (2005-02-15)
The cytoprotective drug amifostine (Ethyol) protects rats from oral mucositis resulting from a single dose of gamma-irradiation. We expanded earlier studies to determine whether multiple doses of amifostine protect against fractionated or hyperfractionated radiation and whether the active metabolite of
Jeffrey S Murley et al.
Radiation research, 175(1), 57-65 (2010-12-24)
RKO36 cells exposed to either WR1065 or 10 cGy X rays show elevated SOD2 gene expression and SOD2 enzymatic activity. Cells challenged at this time with 2 Gy exhibit enhanced radiation resistance. This phenomenon has been identified as a delayed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service