Skip to Content
Merck
All Photos(5)

Documents

A4881

Sigma-Aldrich

DL-Arginine hydrochloride

≥98% (TLC)

Synonym(s):

Arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Arginine hydrochloride, ≥98% (TLC)

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

128-130 °C

solubility

H2O: soluble

SMILES string

Cl.NC(CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H

InChI key

KWTQSFXGGICVPE-UHFFFAOYSA-N

Application

DL-Arginine hydrochloride (DL-Arg) is a racemic mixture of the natural proteinogenic amino acid L-arginine and the non-proteinogenic D-arginine. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.

Substrates

Substrate for nitric oxide synthetase; induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siddhartha Roy et al.
Acta crystallographica. Section B, Structural science, 61(Pt 1), 89-95 (2005-01-22)
The adipic acid complexes of DL-arginine and L-arginine are made up of zwitterionic, singularly positively charged arginium ions and doubly negatively charged adipate ions, with a 2:1 stoichiometry. One of the two crystallographically independent arginium ions in the L-arginine complex
N T Saraswathi et al.
Acta crystallographica. Section B, Structural science, 59(Pt 5), 641-646 (2003-10-31)
The complexes of L-arginine and DL-lysine with pimelic acid are made up of singly positively charged zwitterionic amino acid cations and doubly negatively charged pimelate ions in a 2:1 ratio. In both structures, the amino acid molecules form twofold symmetric
Yosuke Nakano et al.
Journal of bioscience and bioengineering, 123(1), 134-138 (2016-08-21)
d-Amino acids have recently attracted much attention in various research fields including medical, clinical and food industry due to their important biological functions that differ from l-amino acid. Most chiral amino acid separation techniques require complicated derivatization procedures in order
Kohei Yoshikawa et al.
Journal of bioscience and bioengineering, 130(4), 437-442 (2020-07-04)
Fast enantiomeric separation of amino acids was studied by liquid chromatography/mass spectrometry (LC/MS) on a chiral crown ether stationary phase. A chiral crown ether bonded silica column (3 mm internal diameter (i.d.), 5 cm long) packed with 3 μm particles was employed instead
Moyu Taniguchi et al.
Journal of bioscience and bioengineering, 124(4), 414-418 (2017-06-02)
Although naturally abundant amino acids are represented mainly by l-enantiomers, fermented foods are known to contain various d-amino acids. Enantiospecific profiles of food products can vary due to fermentation by bacteria, and such alterations may contribute to changes in food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service