Skip to Content
Merck
All Photos(1)

Key Documents

A1270000

Arginine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Arginine, (S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

arginine

manufacturer/tradename

EDQM

mp

222 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Arginine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jurgen R Piet et al.
The Journal of infectious diseases, 209(11), 1781-1791 (2013-12-18)
Streptococcus pneumoniae (pneumococcus) is a major human pathogen causing pneumonia, sepsis and bacterial meningitis. Using a clinical phenotype based approach with bacterial whole-genome sequencing we identified pneumococcal arginine biosynthesis genes to be associated with outcome in patients with pneumococcal meningitis.
Lindsay D Plank et al.
Hepatology (Baltimore, Md.), 61(2), 639-647 (2014-09-13)
Preliminary work suggested that perioperative immunonutrition (IMN) enriched in n-3 fatty acids, arginine, and nucleotides may improve preoperative nutritional status, enhance postoperative recovery, and reduce postoperative infectious complications in patients undergoing liver transplantation (LT). The current study examined these outcomes
Phyllis S Y Chong et al.
Cancer research, 74(11), 3043-3053 (2014-04-02)
PRL-3, an oncogenic dual-specificity phosphatase, is overexpressed in 50% of acute myelogenous leukemia (AML) and associated with poor survival. We found that stable expression of PRL-3 confers cytokine independence and growth advantage of AML cells. However, how PRL-3 mediates these
Kaoru Kumazaki et al.
Nature, 509(7501), 516-520 (2014-04-18)
Newly synthesized membrane proteins must be accurately inserted into the membrane, folded and assembled for proper functioning. The protein YidC inserts its substrates into the membrane, thereby facilitating membrane protein assembly in bacteria; the homologous proteins Oxa1 and Alb3 have
Yannick Auclair et al.
DNA repair, 12(7), 459-465 (2013-05-21)
Post-translational modifications are well-known modulators of DNA damage signaling and epigenetic gene expression. Protein arginine methylation is a covalent modification that results in the addition of methyl groups to the nitrogen atoms of the arginine side chains and is catalyzed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service