Skip to Content
Merck
All Photos(1)

Key Documents

A4290

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) F(ab′)2 fragment−Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Goat polyclonal anti-Human IgM (μ-chain specific) F(ab′)2 fragment−Peroxidase antibody is specific for human IgM when tested against purified human IgA, IgG, IgM, Bence Jones Kappa and Bence Jones Lambda myeloma proteins.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM antibodies are responsible for agglutination of red blood cells in mis-matched blood transfusions. The level of IgM may vary with the status of disease or infection.
Horseradish Peroxidase (HRP) is an enzyme that catalyzes the conversion of chromogenic substrates such as o-phenylenediamine (OPD), 4-chloro-1-naphthol 3,3′,5,5′-tetramethylbenzidine (TMB), 3,3′-Diaminobenzidine (DAB) or 2,2′-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) (ABTS); chemiluminescent substrates such as CPS-3 (enhanced luminal) and fluorogenic substrates such as Ampliflu Red into detectable chromophores, light-emitters or fluorescers, respectively.

Immunogen

purified human IgM

Application

Goat polyclonal anti-Human IgM (μ-chain specific) F(ab′)2 fragment−Peroxidase antibody may be used to detect and quantitate the level of IgM in human serum and biological fluids via chromogenic, chemoluminescent or fluorogenic immunochemical or immunohistochemical techniques. ELISA assays on EBV +BCLs established from EBV+ human B cells were performed using HRP conjugated goat anti-human IgM mu chain specific as the secondary. Secondary antibody was incubated for 2 hrs at room temperature.
Peroxidise-conjugated goat F(ab′)2 fragment anti-human IgM (μ-chain specific) has been used as a secondary antibody to detect human IgMs to hantanvirus antigens using ELISA.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced FVIII recovery associated with anti-FVIII PEG antibodies after BNT162b2 SARS-CoV-2 vaccination.
Carla Valsecchi et al.
Blood advances, 7(1), 174-177 (2022-10-29)
T Avsic-Zupanc et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 28(4), 860-865 (2000-05-29)
This report describes the first investigation of clinical findings for a larger series of patients with hemorrhagic fever with renal syndrome (HFRS) who were infected with Dobrava virus. From 1985 to 1995, 38 patients with serologically confirmed HFRS were hospitalized
F Elgh et al.
Journal of clinical microbiology, 35(5), 1122-1130 (1997-05-01)
Worldwide, hantaviruses cause more than 100,000 human infections annually. Rapid and accurate methods are important both in monitoring acute infections and for epidemiological studies. We and others have shown that the amino termini of hantavirus nucleocapsid proteins (Ns) are sensitive
Danijela Konforte et al.
Journal of immunology (Baltimore, Md. : 1950), 177(12), 8381-8392 (2006-12-05)
The complex process of B cell development is controlled by multiple factors from the surrounding microenvironment including cytokines. IL-21 is a recently identified type I cytokine, mainly produced by activated CD4(+) T cells. It has been shown to promote differentiation
Jaspreet Jain et al.
The American journal of tropical medicine and hygiene, 102(4), 857-868 (2020-02-19)
Chikungunya fever (CHIKF) is a major public health concern and is caused by chikungunya virus (CHIKV). In 2005, the virus was reintroduced into India, resulting in massive outbreaks in several parts of the country. During 2010 and 2016 outbreaks, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service