Skip to Content
Merck
All Photos(1)

Key Documents

I0884

Sigma-Aldrich

Anti-Human IgA (α-chain specific) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti Human IgA Antibody - Anti-Human IgA (α-chain specific) antibody produced in goat, Anti Human Iga

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgA antibody is secretory antibody and is present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Thus secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces
Anti-Human IgA (α-chain specific) antibody specifically recognizes human IgA.
Immunoglobulin A (IgA) is the primary antibody involved in mucosal immunity. It is identified by the presence of α-type heavy chains. IgA which is composed of two subclasses IgA1 and IgA2 can exist in a dimeric secretory form found in mucous secretions.
Specificity for the α-chain of human IgA is determined by Ouchterlony Double Diffusion (ODD). The antibody preparation is specific for human IgA when tested against purified human IgA, IgG, IgM, Bence Jones kappa and Bence Jones λ myeloma proteins.

Immunogen

Purified human IgA

Application

Anti-Human IgA (α-chain specific) antibody is suitable for Ouchterlony double diffusion. The antibody was used for IgA quantification by ELISA in various studies. Antibody dilution of 1:4000 was used for passive hemagglutination inhibition assay.
Goat polyclonal anti-human IgA (α-chain specific) antibody may be used to locate and identify the α-chain of human IgA by immunocytochemical and immunohistochemical (IHC) techniques.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leif K Nielsen et al.
Transfusion, 48(9), 1892-1897 (2008-06-03)
To avoid anaphylactic reactions, immunoglobulin (Ig)A-deficient patients with anti-IgA should be transfused with IgA-deficient blood components. There is a need for fast and robust assays for demonstration of IgA deficiency and for detection of anti-IgA. Recombinant human IgA1 and IgA2
Mikael Sonesson et al.
European journal of oral sciences, 119(1), 15-20 (2011-01-20)
According to previous studies, minor glands produce about 35% of the total salivary immunoglobulin A (salivary IgA). The age-dependent increase in whole-saliva salivary IgA concentrations has been studied extensively, but we found no published reports comparing the minor-gland saliva concentrations
Hannah Andersson et al.
International journal of microbiology, 2016, 8469018-8469018 (2017-01-20)
Objective. To clarify the effect of Lactobacillus plantarum 299v on the salivary cortisol and salivary IgA levels in young adults under examination stress. Design. Forty-one students with an upcoming academic exam were included in a randomized double-blind, placebo-controlled study. The
Dan Ericson et al.
Pathogens and disease, 68(3), 82-87 (2013-06-06)
We investigated whether ingestion of probiotic bacteria could influence salivary IgA levels, specific anti-mutans streptococci IgA levels and specific antibodies towards the ingested probiotic bacterium. The study was a randomised, double-blind, placebo-controlled trial, where the test group (n = 11) received twice
P Macchiaverni et al.
Scandinavian journal of immunology, 74(6), 619-627 (2011-09-03)
There is strong evidence from animal models that placental and/or breast milk-mediated transfer of maternal allergen-specific IgG prevents allergic immune responses in the progeny. Both human and animal data also point to IgA as having an important regulatory role. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service