Skip to Content
Merck
All Photos(3)

Key Documents

A4062

Sigma-Aldrich

Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Rabbit Anti-Goat IgG (whole molecule)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

goat

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulins (Igs) belongs immunoglobulin super-family. There are five different types of Immunoglobulins. Each immunoglobin have two heavy (H) and two light (L), held together by disulphide linkages. Each light comprises of one variable N-terminal region and a constant C-terminal region. Heavy chain has one variable N-terminal region and three or four constant (CH1-CH4) C-terminal region.

Immunogen

purified goat IgG

Application

Alkaline phosphatase conjugated rabbit anti goat IgG was used as the secondary for ELISA assays to detect the binding of bacterial proteins to C components. The antibody was used at 1:5000 dilutions and detected using P-nitrophenyl phosphate at 1mg/ml (Sigma) as the substrate.
Anti-Goat IgG (whole molecule)-alkaline phosphatase antibody can be used in western blotting and immunoblotting (diluted 1: 20,000) .

Biochem/physiol Actions

IgG antibodies regulate several functions such as complement activation and phagocytosis. Thus, they play a crucial role in facilitating cytological immune responses. Anti-goat IgG (whole molecule)-alkaline phosphatase antibody (diluted 1: 5000) can be used for anti-OVA IgG detection. Rabbit anti- goat IgG (whole molecule)-alkaline phosphatase antibody reacts specifically with all goat immunoglobulins.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 10 mM glycine, 1 mM MgCl2, 50% glycerol and 15 mM sodium azide

Preparation Note

Adsorbed to reduce background staining with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Pausa et al.
Journal of immunology (Baltimore, Md. : 1950), 170(6), 3214-3222 (2003-03-11)
Borrelia burgdorferi, the etiological agent of Lyme disease, comprises three genospecies, Borrelia garinii, afzelii, and burgdorferi sensu strictu, that exhibit different pathogenicity and differ in the susceptibility to C-mediated killing. We examined C-sensitive and C-resistant strains of B. burgdorferi for
Xinyan Wang et al.
Journal of molecular histology, 52(4), 741-749 (2021-05-29)
Accumulating evidence has suggests that women with advanced endometriosis exhibit alterations in the expression of genes in the endometrium compared to healthy controls. Furthermore, replication stress is a characteristic feature of cancer cells, which results from sustained proliferative signaling induced
Chun-E Ren et al.
International journal of molecular sciences, 16(3), 5864-5885 (2015-03-18)
Epithelial stromal cells represent a major cellular component of human uterine endometrium that is subject to tight hormonal regulation. Through cell-cell contacts and/or paracrine mechanisms, stromal cells play a significant role in the malignant transformation of epithelial cells. We isolated
Nikita Deshpande et al.
Genetics, 209(4), 1085-1097 (2018-06-21)
Many heterogametic organisms adjust sex chromosome expression to accommodate differences in gene dosage. This requires selective recruitment of regulatory factors to the modulated chromosome. How these factors are localized to a chromosome with requisite accuracy is poorly understood. Drosophila melanogaster
S Nutten et al.
Clinical and vaccine immunology : CVI, 14(2), 190-197 (2006-12-08)
In this study, we assessed the effect of administering the antibiotic amoxicillin to rat pups on the immune response to orally fed ovalbumin (OVA). We first established that amoxicillin administration durably altered the gut microbiota of these animals. In parallel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service