Skip to Content
Merck
All Photos(1)

Key Documents

F4262

Sigma-Aldrich

Anti-Cat IgG (whole molecule)−FITC antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Cat IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

FITC conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:32

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG is present in large quantities in the human serum. It constitutes about 10-20% of the plasma proteins. IgG is composed of glycoproteins, out of which it is 82-96% proteins and 4-18% carbohydrates. It consists of four sub-classes i.e IgG1, IgG2, IgG3, and IgG4. IgG is composed of four polypeptide chains-two heavy chains (γ chains) and two light chains (κ or λ chains) which are linked by inter-chain disulfide bonds. The heavy chains consist of a N-terminal variable domain (VH) and three constant domains (CH1, CH2, CH3). A hinge region exists between the CH1 and CH2 region. The light chains have one N-terminal variable domain (VL) and one constant domain (CL). The heavy and the light chains are linked at VH and CH1 domain to form the Fab arm (Fragment antigen binding). The antigen binds to the V regions of the antibody.

Specificity

Anti-Cat IgG (whole molecule)-FITC antibody specifically recognizes cat IgG. Goat anti-cat IgG is conjugated to Fluorescein Isothiocyanate (FITC) in an alkaline reaction, then further purified to remove unbound FITC.

Immunogen

Purified cat IgG
Purified cat IgG.

Application

Anti-Cat IgG (whole molecule)-FITC antibody has been used in immunofluorescence antibody test (IFAT), immunofluorescence.
Anti-Cat IgG (whole molecule)-FITC antibody may be used for immunofluorescence at a working antibody dilution of 1:32. The antibody was used in indirect fluorescent antibody test (IFAT) to detect Toxoplasma specific antibodies in cat at a dilution of 1:100.
Anti-cat IgG (whole molecule) −FITC antibody can be used in direct (1:32) and indirect immunofluorescence (dilution 1:64).

Biochem/physiol Actions

IgG antibody plays a crucial role in humoral immune responses such as phagocytosis, complement activation, placental transport and cell surface-receptor binding. FITC (fluorescein isothiocyanate), a fluorochrome dye conjugation is useful in localization of antigen-antibody interaction of biological structures like cells or tissues. Anti-cat IgG (whole molecule) -FITC antibody can be used in immunofluorescence study of exfoliative dermatitis in cats. Goat anti-cat IgG antibody is immonospecific for cat IgG.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Mercedes Nogueras et al.
PloS one, 8(8), e71386-e71386 (2013-08-14)
Rickettsiatyphi is the etiological agent of murine typhus (MT), a disease transmitted by two cycles: rat-flea-rat, and peridomestic cycle. Murine typhus is often misdiagnosed and underreported. A correct diagnosis is important because MT can cause severe illness and death. Our
S Rottenberg et al.
Veterinary pathology, 41(4), 429-433 (2004-07-03)
Five cases of exfoliative dermatitis in cats were presented from 1996 to 2002 in which a feline thymoma was found by postmortem or postsurgical examination. Besides abundant exfoliation of keratin squames and layers, the histologic picture of the skin revealed
Gabriela Hartmann et al.
Parasitology research, 122(11), 2631-2639 (2023-09-05)
Feline leishmanial infection is reported worldwide, but the epidemiological role of domestic cats in the leishmaniasis cycle remains unclear, and cats might act as cryptic reservoir hosts in endemic areas with no feline leishmaniosis cases. Considering that, a serological screening
Exploring the epidemiological role of the Eurasian lynx (Lynx lynx) in the life cycle of Toxoplasma gondii.
Scherrer, et al.
International Journal for Parasitology. Parasites and Wildlife, 21, 10-Jan-10-Jan (2023)
Feline Leishmania infection in a canine leishmaniasis endemic region, Portugal
Maia C, et al.
Veterinary Parasitology, 174(3-4), 336-340 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service