Skip to Content
Merck
All Photos(1)

Key Documents

A4003

Sigma-Aldrich

Ala-Tyr

≥98% (TLC), suitable for mass spectrometry (MS)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O4
CAS Number:
Molecular Weight:
252.27
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

Ala-Tyr,

Assay

≥98% (TLC)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(O)=O

InChI

1S/C12H16N2O4/c1-7(13)11(16)14-10(12(17)18)6-8-2-4-9(15)5-3-8/h2-5,7,10,15H,6,13H2,1H3,(H,14,16)(H,17,18)/t7-,10-/m0/s1

InChI key

ALZVPLKYDKJKQU-XVKPBYJWSA-N

Gene Information

human ... SLC15A1(6564)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Ala-Tyr

Biochem/physiol Actions

Alanyl dipeptides such as ala-leu, ala-lys, ala-gly, ala-pro, ala-tyr and ala-phe may be used in physicochemical studies or to evaluate dipeptide separation technologies. Alanyl dipeptides may also be used for studying cell uptake mechanisms, dipeptide metabolism or cell growth supplementation benefits.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yanjun Yang et al.
Journal of agricultural and food chemistry, 55(19), 7891-7895 (2007-08-25)
Corn gluten meal (CGM) was hydrolyzed by Alcalase after starch removal of CGM was applied as a pretreatment. A new inhibitory peptide for angiotensin I-converting enzyme (ACE) was isolated from the hydrolysate of CGM with the use of Bio-Rad P-2
Joanna Tkaczewska et al.
Molecules (Basel, Switzerland), 24(1) (2019-01-02)
The protein by-products from carp (Cyprinus carpio) are normally discarded as industrial waste during fish processing. The objective of this study was to identify and characterise the peptides with a potential antioxidant activity that are released from carp skin proteins
K Meylaers et al.
Current pharmaceutical design, 9(2), 159-174 (2003-02-07)
The number of bacterial and fungal strains that have developed resistance against the classical antibiotics continues to grow. The intensified search for new antibiotic lead compounds has resulted in the discovery of numerous endogenous peptides with antimicrobial properties in plants
Hancheng Cai et al.
Dalton transactions (Cambridge, England : 2003), (27)(27), 5395-5400 (2009-07-01)
Copper-64 shows promise as both a suitable PET imaging and therapeutic radionuclide due to its nuclear characteristics. Stable attachment of radioactive (64)Cu(2+) to targeted imaging probes requires the use of a bifunctional chelator. Sarcophagine (Sar) ligands coordinate the metal ion
Lihui Wei et al.
Nuclear medicine and biology, 36(3), 277-285 (2009-03-28)
The alpha(v)beta(3) integrin is a cell adhesion molecule known to be involved in stages of angiogenesis and metastasis. In this study, the chelators CB-TE2A and diamsar were conjugated to cyclic RGDyK and RGDfD and the biological properties of (64)Cu-labeled peptides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service