Skip to Content
Merck
All Photos(1)

Key Documents

A8052

Sigma-Aldrich

Arg-Gly-Asp

≥97% (TLC)

Synonym(s):

L-arginyl-glycyl-L-aspartic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22N6O6
CAS Number:
Molecular Weight:
346.34
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic

Assay

≥97% (TLC)

form

powder

technique(s)

cell culture | stem cell: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C12H22N6O6/c13-6(2-1-3-16-12(14)15)10(22)17-5-8(19)18-7(11(23)24)4-9(20)21/h6-7H,1-5,13H2,(H,17,22)(H,18,19)(H,20,21)(H,23,24)(H4,14,15,16)/t6-,7-/m0/s1

InChI key

IYMAXBFPHPZYIK-BQBZGAKWSA-N

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Arg-Gly-Asp

General description

Arg-Gly-Asp (RGD) is an integrin binding site, which belongs to the class of adhesive proteins. It functions as a brain tumor targeting ligand.

Application

Arg-Gly-Asp has been used:
  • to saturate the cell surface Arg-Gly-Asp (RGD) receptors and inhibit marrow stromal cell (MSC) attachment to peptide modified hydrogel
  • in adhesion assay
  • in inhibition assay to confirm the β1-integrin-dependency of cell adhesion
  • in textile fabrication and RGD textile functionalization
  • to engineer a population of human dermal fibroblasts with impaired migration
  • to modify glass substrate and evaluate the behavior of human bone marrow mesenchymal stem cells (hBM-MSCs)
  • in the cell culture of human mammary epithelial cells

Biochem/physiol Actions

Primary sequence involved with the binding of proteins to cell surfaces

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Tripeptide from the cell-attachment domain of fibronectin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of incorporating RGD adhesive peptide in polyethylene glycol diacrylate hydrogel on osteogenesis of bone marrow stromal cells
Yang F, et al.
Biomaterials, 26(30), 5991-5998 (2005)
Evaluation of the Osteoblast Behavior to PGA Textile Functionalized with RGD as a Scaffold for Bone Regeneration
Ortiz M, et al.
Journal of Nanomaterials, 2017 (2017)
Bio-active molecules modified surfaces enhanced mesenchymal stem cell adhesion and proliferation
Mobasseri R, et al.
Biochemical and biophysical research communications, 483(1), 312-317 (2017)
Cell attachment to thrombospondin: the role of ARG-GLY-ASP, calcium, and integrin receptors.
Lawler J, et al.
The Journal of cell biology, 107(6), 2351-2361 (1988)
Engineering the migration and attachment behaviour of primary dermal fibroblasts
Garcia-Gareta E, et al.
Biotechnology and Bioengineering (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service