Skip to Content
Merck
All Photos(1)

Key Documents

A2736

Sigma-Aldrich

Anastrozole

≥98% (HPLC)

Synonym(s):

2;2"-[5-(1H-1;2;4-Triazol-1-ylmethyl)-1, 3-Phenylene]bis(2-methyl-propiononitrile, Arimidex, ICI-D1033, ZD1033, a,a,a′,a′-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N5
CAS Number:
Molecular Weight:
293.37
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 40 mg/mL

originator

AstraZeneca

storage temp.

room temp

SMILES string

[n]2(ncnc2)Cc1cc(cc(c1)C(C)(C)C#N)C(C)(C)C#N

InChI

1S/C17H19N5/c1-16(2,9-18)14-5-13(8-22-12-20-11-21-22)6-15(7-14)17(3,4)10-19/h5-7,11-12H,8H2,1-4H3

InChI key

YBBLVLTVTVSKRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anastrozole (aromatase inhibitor) has been used:
  • as a positive control in DNA fragmentation (ladder) assay
  • to investigate its effects along with extra virgin olive oil and its major fatty acid component (omega-9 OA) in estrogen receptor positive mammary adenocarcinoma cells
  • to study its effects on viability, cell proliferation and apoptosis in Glioblastoma multiforme model in vivo

Biochem/physiol Actions

Anastrozole is a nonsteroidal aromatase inhibitor.
Anastrozole, which contains a triazole functional group, reversibly binds to the cytochrome P-450 component of aromatase. Binding interferes with the catalytic properties of aromatase, which results in inhibition of estrogen synthesis.
The aromatase enzyme converts adrenal androgens to estrogen; this enzymatic activity is the primary source of estrogen production in postmenopausal women. One treatment for estrogen receptor-positive breast cancer in postmenopausal women is through inhibition of aromatase. Anastrozole is a nonsteroidal, benzyl-triazole derivative that inhibits aromatase through competitive inhibition and is used to treat estrogen receptor-positive breast cancer. This compound is considered a third-generation, Type II aromatase inhibitor because it is more selective and less effective (if at all) on other steroidal hormones than first and second generation inhibitors.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anastrozole Reduce Cell Proliferation and Induce Apoptosis in Glioblastoma Multiforme Xenograft Mouse Model
Aguilar G, et al.
Journal of Cancer Science & Therapy, 9(3), 655-660 (2017)
Dennis C Sgroi et al.
The Lancet. Oncology, 14(11), 1067-1076 (2013-09-17)
Biomarkers to improve the risk-benefit of extended adjuvant endocrine therapy for late recurrence in patients with oestrogen-receptor-positive breast cancer would be clinically valuable. We compared the prognostic ability of the breast-cancer index (BCI) assay, 21-gene recurrence score (Oncotype DX), and
Extra virgin olive oil potentiates the effects of aromatase inhibitors via glutathione depletion in estrogen receptor-positive human breast cancer (MCF-7) cells
Ismail AM, et al.
Food And Chemical Toxicology, 62(3), 817-824 (2013)
Irene Guadalupe Aguilar-García et al.
Brain sciences, 13(3) (2023-03-30)
Glioblastoma Multiforme (GBM) is a tumor that infiltrates several brain structures. GBM is associated with abnormal motor activities resulting in impaired mobility, producing a loss of functional motor independence. We used a GBM xenograft implanted in the striatum to analyze
P I Petkov et al.
SAR and QSAR in environmental research, 20(7-8), 657-678 (2009-12-22)
Cytochrome P450 aromatase is a key steroidogenic enzyme that converts androgens to estrogens in vertebrates. There is much interest in aromatase inhibitors (AIs) both because of their use as pharmaceuticals in the treatment of estrogen-sensitive breast cancers, and because a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service