Skip to Content
Merck
All Photos(1)

Key Documents

Y0001522

Anastrozole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2;2"-[5-(1H-1;2;4-Triazol-1-ylmethyl)-1, 3-Phenylene]bis(2-methyl-propiononitrile, Arimidex, ICI-D1033, ZD1033, a,a,a′,a′-Tetramethyl-5-(1H-1,2,4-triazol-1-ylmethyl)-1,3-benzenediacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N5
CAS Number:
Molecular Weight:
293.37
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

anastrozole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[n]2(ncnc2)Cc1cc(cc(c1)C(C)(C)C#N)C(C)(C)C#N

InChI

1S/C17H19N5/c1-16(2,9-18)14-5-13(8-22-12-20-11-21-22)6-15(7-14)17(3,4)10-19/h5-7,11-12H,8H2,1-4H3

InChI key

YBBLVLTVTVSKRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Anastrozole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anastrozole is a nonsteroidal aromatase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Howell et al.
Breast cancer research and treatment, 82(3), 215-222 (2004-01-02)
Tumors that have spread to the liver or lungs (visceral metastases) are associated with a worse prognosis than tumors in soft tissue and bone only. Here we review available efficacy data to address whether or not anastrozole, a non-steroidal aromatase
Catherine M Kelly et al.
Expert opinion on drug safety, 9(6), 995-1003 (2010-10-07)
Anastrozole is a third-generation aromatase inhibitor used in the adjuvant setting for the treatment of hormone receptor-positive breast cancer. Several adjuvant randomized trials have reported greater efficacy for anastrozole when compared to tamoxifen. This review discusses the mechanism of action
Tuna Yucel et al.
Biomaterials, 35(30), 8613-8620 (2014-07-11)
A silk-protein based reservoir rod was developed for zero-order and long-term sustained drug delivery applications. Silk reservoir rod formulations were processed in three steps. First, a regenerated silk fibroin solution, rich in random-coil content was transformed into a tubular silk
Peter H Graham
Expert opinion on pharmacotherapy, 8(14), 2347-2357 (2007-10-12)
Anastrozole is a third-generation aromatase inhibitor with an extensive evidence base to preferentially support its use in multiple scenarios in endocrine-responsive breast cancer. These include neoadjuvant treatment, immediate and delayed adjuvant treatment, salvage treatment and first-line treatment of advanced breast
Raimund Jakesz
Expert opinion on drug metabolism & toxicology, 2(2), 301-312 (2006-07-27)
Anastrozole is a nonsteroidal, third-generation aromatase inhibitor, which is heralded as an effective alternative endocrine therapy to tamoxifen in postmenopausal women with hormone-responsive breast cancer. Anastrozole has a high affinity for aromatase, a CYP enzyme involved in estrogen synthesis, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service