Skip to Content
Merck
All Photos(1)

Key Documents

A1970

Millipore

Anti-VSV-Glycoprotein−Agarose antibody, Mouse monoclonal

clone P5D4, purified from hybridoma cell culture, PBS suspension

Synonym(s):

Monoclonal Anti-VSV Glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P5D4, monoclonal

form

PBS suspension

analyte chemical class(es)

proteins (VSV-G)

technique(s)

immunoprecipitation (IP): suitable
protein purification: suitable

isotype

IgG1

capacity

≥15 nmol/mL, resin binding capacity (VSV-G tagged fusion protein)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Anti-VSV-G Agarose Conjugate is the immunoglobulin fraction of Monoclonal Anti-VSV glycoprotein covalently linked to agarose.

Specificity

The antibody recognizes an epitope containing the five carboxy-terminal amino acids of VSV Glycoprotein. In infected cells, the antibody localizes the immature forms of VSV-G in the rough endoplasmic reticulum (RER) and in the cisternae of Golgi complex, as well as mature VSV-G at the cell surface and in the budding virus. The antibody does not stain the secreted form of VSV-G which lacks the membrane and the cytoplasmic domain. This antibody has been used for studies on the role of the cytoplasmic domain on newly-synthesized VSV-G during transfer to the plasma membrane and cell surface, using micro-injected antibody, immunoblotting, immunoprecipitation, immunocytochemistry and immunoelectron microscopy. The antibody has been used for the detection, immunoprecipitation and immunocytochemical staining of exogenously introduced constructs tagged with the carboxyl-terminus of VSV-G. This tag does not interfere with the function of the studied protein and can be specifically recognized by the P5D4 antibody without cross-reaction with any endogenous protein.

Immunogen

synthetic peptide containing the 15 carboxy-terminal amino acids (497-511) of Vesicular Stomatitis Virus Glycoprotein (VSV-G), conjugated to KLH.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)
For immunoprecipitation and affinity purification of VSV-G tagged fusion proteins. The antibody recognizes an epitope containing the five carboxy-terminal amino acids of VSV Glycoprotein.

Physical form

Supplied as a suspension (1:1,v/v) of beaded agarose in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Preparation Note

Prepared using cyanogen bromide-activated agarose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew M Lippa et al.
Molecular microbiology, 115(6), 1138-1151 (2020-11-28)
The H-NS-like proteins MvaT and MvaU act coordinately as global repressors in Pseudomonas aeruginosa by binding to AT-rich regions of the chromosome. Although cells can tolerate loss of either protein, identifying their combined regulatory effects has been challenging because the
Yael Katan-Khaykovich et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(4), 1296-1301 (2011-01-12)
Nucleosome deposition occurs on newly synthesized DNA during DNA replication and on transcriptionally active genes via nucleosome-remodeling complexes recruited by activator proteins and elongating RNA polymerase II. It has been long believed that histone deposition involves stable H3-H4 tetramers, such
Sachin Mohan et al.
The Journal of biological chemistry, 285(45), 34566-34578 (2010-08-26)
The small intestinal BB Na(+)/H(+) antiporter NHE3 accounts for the majority of intestinal sodium and water absorption. It is highly regulated with both postprandial inhibition and stimulation sequentially occurring. Phosphatidylinositide 4,5-bisphosphate (PI(4,5)P(2)) and phosphatidylinositide 3,4,5-trisphosphate (PI(3,4,5)P(3)) binding is involved with
Bridget R Kulasekara et al.
eLife, 2, e01402-e01402 (2013-12-19)
Individual cell heterogeneity is commonly observed within populations, although its molecular basis is largely unknown. Previously, using FRET-based microscopy, we observed heterogeneity in cellular c-di-GMP levels. In this study, we show that c-di-GMP heterogeneity in Pseudomonas aeruginosa is promoted by
Larry A Gallagher et al.
Nature microbiology, 7(6), 844-855 (2022-06-02)
DNA-protein interactions are central to fundamental cellular processes, yet widely implemented technologies for measuring these interactions on a genome scale in bacteria are laborious and capture only a snapshot of binding events. We devised a facile method for mapping DNA-protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service