Skip to Content
Merck
All Photos(3)

Key Documents

A1805

Sigma-Aldrich

ACCUSPIN Tubes Sterile, 12 mL Capacity

radiation sterilized tube fitted with a high density polyethylene barrier

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

sterility

sterile; irradiated

form

tubes

application(s)

hematology
histology

storage temp.

15-25°C

General description

ACCUSPIN™ Tubes are polypropylene radiation sterilized tube fitted with a high density polyethylene barrier. Each tube will accept 3mL of the density gradient.

Application

ACCUSPIN™ Tubes Sterile, 12 mL Capacity is intended for use with the density gradient separation medium Histopaque™ -1077:

  • in the isolation of lymphocytes and other mononuclear cells.
  • isolation of cord blood–derived endothelial progenitor cells.citation
  • isolate mononuclear cell layer from bone marrow samples.
  • suitable for the purification of human bone marrow aspirates for the investigation of the therapeutic value of BET inhibitors for acute myeloid leukemia.
  • isolation of cord blood-derived endothelial progenitor cells

Principle

Following the addition of Histopaque-1077 to the ACCUSPIN Tube, a brief centrifugation places the Histopaque-1077 below the frit. The blood sample can be added to the top chamber of the tube without the risk of mixing with the Histopaque-1077 in the lower chamber under the frit. On subsequent centrifugation the whole blood descends through the frit to contact with theHistopaque-1077. The elements of greater density displace a volume of Histopaque-1077 above the forgiving a clear separation of the blood components. The erythrocytes aggregate and the granulocytes become lightly hypertonic, increasing their sedimentation rate, resulting in pelleting at the bottom of the ACCUSPIN Tube. Lymphocytes and other mononuclear cells, e.g., monocytes, remain at the plasma/Histopaque-1077interface. This dense band of mononuclear cells maybe collected by pouring off the contents of the upper chamber or by means of a pipette. Erythrocyte contamination is avoided due to the barrier between the chambers.

Quantity

Package contains 20 tubes

Legal Information

Accuspin is a trademark of Sigma-Aldrich Co. LLC
Histopaque is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service