Skip to Content
Merck
All Photos(1)

Key Documents

A0966

Sigma-Aldrich

4-Amino-1,8-naphthalimide

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O2
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

4-Amino-1,8-naphthalimide,

form

solid

mp

360 °C

density

1.105 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Nc1ccc2C(=O)NC(=O)c3cccc1c23

InChI

1S/C12H8N2O2/c13-9-5-4-8-10-6(9)2-1-3-7(10)11(15)14-12(8)16/h1-5H,13H2,(H,14,15,16)

InChI key

SSMIFVHARFVINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

4-Amino-1,8-naphthalimide sensitizes cells to radiation-induced cell damage and enhances the cytotoxicity of 1-methyl-3-nitro-1-nitrosoguanidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Banasik et al.
The Journal of biological chemistry, 267(3), 1569-1575 (1992-01-25)
Two classes of enzymes, poly(ADP-ribose) synthetase and mono(ADP-ribosyl)transferases, catalyze covalent attachment of multiple or single residues, respectively, of the ADP-ribose moiety of NAD+ to various proteins. In order to find good inhibitors of poly(ADP-ribose) synthetase free of side actions and
A Schlicker et al.
International journal of radiation biology, 75(1), 91-100 (1999-02-11)
Poly(ADP-ribose) polymerase (PARP; EC 2.4.2.30) is a chromatin-bound enzyme which is known to regulate chromatin structure by poly(ADP-ribosyl)ation of nuclear proteins, to facilitate DNA base excision repair, and to contribute to cellular recovery following DNA damage. Because inhibitors of PARP
Marco A Alcala et al.
Nanomedicine : nanotechnology, biology, and medicine, 7(3), 249-258 (2010-10-16)
Surgery is currently the best approach for treating either primary or metastatic hepatic malignancies. Because only 20% of hepatic cancers are operable in patients, several types of regional therapy (RT) are emerging as alternate treatment modalities. However, RTs can have
Emma B Veale et al.
The Journal of organic chemistry, 75(16), 5513-5525 (2010-08-14)
The synthesis and characterization of three bis-1,8-naphthalimide-containing Tröger's bases 1-3, formed from the corresponding 4-amino-1,8-naphthalimide precursors 7-9 in a single step, is described. The photophysical investigation of 1-3 and 7-9 was carried out in various organic solvents as well as
Jun Feng Zhang et al.
Organic letters, 13(5), 1190-1193 (2011-02-09)
A naphthalimide-based highly selective colorimetric and ratiometric fluorescent probe for the fluoride ion displayed both one- and two-photon ratiometric changes. Upon reaction with the F(-) (TBA(+) and Na(+) salts) anion in CH(3)CN as well as in aqueous buffer solution, probe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service