Skip to Content
Merck
All Photos(1)

Key Documents

96631

Sigma-Aldrich

Bis(2,2′-bipyridine)-4′-methyl-4-carboxybipyridine-ruthenium N-succinimidyl ester-bis(hexafluorophosphate)

BioReagent, suitable for fluorescence

Synonym(s):

Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H29F12N7O4P2Ru
CAS Number:
Molecular Weight:
1014.66
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

form

powder

fluorescence

λex 458 nm; λem 628 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Ru++].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.c1ccc(nc1)-c2ccccn2.c3ccc(nc3)-c4ccccn4.Cc5ccnc(c5)-c6cc(ccn6)C(=O)ON7C(=O)CCC7=O

InChI

1S/C16H13N3O4.2C10H8N2.2F6P.Ru/c1-10-4-6-17-12(8-10)13-9-11(5-7-18-13)16(22)23-19-14(20)2-3-15(19)21;2*1-3-7-11-9(5-1)10-6-2-4-8-12-10;2*1-7(2,3,4,5)6;/h4-9H,2-3H2,1H3;2*1-8H;;;/q;;;2*-1;+2

InChI key

UQJZJWDQEYXDCR-UHFFFAOYSA-N

Application

Activated ester of ruthenium complex for acylation of amino acid side chain amines.
Bis(2,2′-bipyridine)-4′-methyl-4-carboxybipyridine-ruthenium N-succinimidyl ester-bis(hexafluorophosphate) has been used:
  • as a component in the preparation of ruthenium labelling agent solution (RuC) as an extrinsic fluorophore
  • to stain protein in mass cytometry
  • in the synthesis of ruthenium (Ru) gelatin
  • in the preparation of streptavidin-Ru(bpy)32+ mixture involved in aptamer-magnetic bead electrochemiluminescence (ECL) sandwich assays

Features and Benefits

This dye has a long fluorescence lifetime (near 500 nanoseconds) with high photostability, good water solubility, a lack of dye-dye interactions and large Stokes′ shifts.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernd Geisser et al.
Inorganic chemistry, 38(9), 2030-2037 (2001-10-24)
The pH-dependent photophysical properties of a series of polypyridyl ruthenium-substituted amino acids were investigated by steady-state and time-resolved luminescence spectroscopy. [H(3)N-DAPA(Rub(2)m)-OH](PF(6))(3) (1), [H(3)N-DABA(Rub(2)m)-OH](PF(6))(3) (2), [H(3)N-Orn(Rub(2)m)-OH](PF(6))(3) (3), and [H(3)N-Lys(Rub(2)m)-OH](PF(6))(3) (4) were obtained by formation of an amide link between the omega-NH(2)
B M Peek et al.
International journal of peptide and protein research, 38(2), 114-123 (1991-08-01)
Boc-L-Lysine derivatives and lysine-containing peptides bearing the electron donor 10H-phenothiazine (PTZ) or the redox chromophore tris(2,2'-bipyridine)ruthenium(II) dication ([Rub3]2+, where b is 2,2'-bipyridine) have been synthesized and characterized. SeO2 oxidation (53% yield) of 4,4'-dimethyl-2,2'-bipyridine, Ag2O oxidation (85% yield) of the monoaldehyde
CellCycleTRACER accounts for cell cycle and volume in mass cytometry data
Rapsomaniki M, et al.
Nature Communications, 9(1), 632-632 (2018)
Fluorometric enzymatic autoindicating biosensor for H2O2 determination based on modified catalase
Ortega E, et al.
Biosensors And Bioelectronics, 41, 150-156 (2013)
Zhejian Li et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(2), 195-199 (2018-10-10)
A novel gold nanoprobe for a sensitive and simple determination of a prostate-specific antigen (PSA) was designed on the basis of homogeneous detection and a peptide cleavage reaction. The gold nanoprobe (AuNPs-peptide-Ru1) consisted of a specific peptide tagged with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service