94640
DL-Valine
ReagentPlus®, ≥99.0% (NT)
Synonym(s):
(±)-α-Aminoisovaleric acid, DL-2-Amino-3-methylbutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH(NH2)COOH
CAS Number:
Molecular Weight:
117.15
Beilstein:
506689
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
≥99.0% (NT)
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
mp
295 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
CC(C)C(N)C(O)=O
InChI
1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)
InChI key
KZSNJWFQEVHDMF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic
K Aaboe et al.
Diabetes, obesity & metabolism, 17(1), 74-81 (2014-09-23)
To examine whether 12 weeks of treatment with a dipeptidyl peptidase-4 (DPP-4) inhibitor, sitagliptin, influences the insulin secretion induced by glucose, glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) during a hyperglycaemic clamp in patients with type 2 diabetes (T2DM).
Yan-Mei Ma et al.
Fish & shellfish immunology, 45(2), 414-421 (2015-05-11)
Streptococcus iniae seriously affects the intensive farming of tilapias. Much work has been conducted on prevention and control of S. iniae infection, but little published information on the metabolic response is available in tilapias against the bacterial infection, and no metabolic
Thomas Rydzak et al.
Journal of industrial microbiology & biotechnology, 42(9), 1263-1272 (2015-07-15)
The ability of Clostridium thermocellum to rapidly degrade cellulose and ferment resulting hydrolysis products into ethanol makes it a promising platform organism for cellulosic biofuel production via consolidated bioprocessing. Currently, however, ethanol yield is far below theoretical maximum due to
Yuka Kamei et al.
The Journal of biological chemistry, 289(46), 32081-32093 (2014-10-09)
Age-related damage accumulates and a variety of biological activities and functions deteriorate in senescent cells. However, little is known about when cellular aging behaviors begin and what cellular aging processes change. Previous research demonstrated age-related mRNA changes in budding yeast
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service