Skip to Content
Merck
All Photos(1)

Key Documents

89270

Sigma-Aldrich

Thymidine

≥99.0% (HPLC)

Synonym(s):

1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Beilstein:
89285
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥99.0% (HPLC)

form

powder

optical activity

[α]20/D +19±1°, c = 1% in H2O

mp

185-190 °C
186-188 °C (lit.)

solubility

hot water: 1 g/10 mL

SMILES string

CC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H14N2O5/c1-5-3-12(10(16)11-9(5)15)8-2-6(14)7(4-13)17-8/h3,6-8,13-14H,2,4H2,1H3,(H,11,15,16)/t6-,7+,8+/m0/s1

InChI key

IQFYYKKMVGJFEH-XLPZGREQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thymidine has been used:
  • to culture human fibroblast cell lines.
  • in in vitro wound healing assay.
  • to culture various cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward J Gane et al.
Gastroenterology, 146(1), 138-146 (2013-09-27)
There is a close relationship between chronic hepatitis B virus infection and chronic renal disease. We analyzed changes in renal function using different markers of glomerular filtration rate (GFR) in multiple studies of telbivudine treatment of patients with chronic hepatitis
Anne P Bouvy et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 29(8), 1587-1597 (2014-04-29)
To evaluate the influence of induction therapy on Tregs we investigated their origin, kinetics and function in kidney transplant patients after treatment with T-cell depleting rabbit antithymocyte globulin (rATG) or IL-2 receptor antagonist basiliximab. Flow cytometry was used to study
Yee Suan Poo et al.
PLoS neglected tropical diseases, 8(12), e3354-e3354 (2014-12-05)
The recent epidemic of the arthritogenic alphavirus, chikungunya virus (CHIKV) has prompted a quest to understand the correlates of protection against virus and disease in order to inform development of new interventions. Herein we highlight the propensity of CHIKV infections
Michael V Airola et al.
PloS one, 9(8), e105830-e105830 (2014-08-22)
Acid sphingomyelinase (aSMase) is a human enzyme that catalyzes the hydrolysis of sphingomyelin to generate the bioactive lipid ceramide and phosphocholine. ASMase deficiency is the underlying cause of the genetic diseases Niemann-Pick Type A and B and has been implicated
Helen Chen et al.
Cell cycle (Georgetown, Tex.), 13(14), 2248-2261 (2014-05-31)
Construction of a mitotic spindle requires biochemical pathways to assemble spindle microtubules and structural proteins to organize these microtubules into a bipolar array. Through a complex with dynein, the receptor for hyaluronan-mediated motility (RHAMM) cross-links mitotic microtubules to provide structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service