Merck
  • Spatial regulation of Aurora A activity during mitotic spindle assembly requires RHAMM to correctly localize TPX2.
Spatial regulation of Aurora A activity during mitotic spindle assembly requires RHAMM to correctly localize TPX2.
Cell cycle (Georgetown, Tex.) (2014-05-31)
Helen Chen, Pooja Mohan, Jihong Jiang, Oksana Nemirovsky, Daniel He, Markus C Fleisch, Dieter Niederacher, Linda M Pilarski, C James Lim, Christopher A Maxwell
ABSTRACT
Construction of a mitotic spindle requires biochemical pathways to assemble spindle microtubules and structural proteins to organize these microtubules into a bipolar array. Through a complex with dynein, the receptor for hyaluronan-mediated motility (RHAMM) cross-links mitotic microtubules to provide structural support, maintain spindle integrity, and correctly orient the mitotic spindle. Here, we locate RHAMM to sites of microtubule assembly at centrosomes and non-centrosome sites near kinetochores and demonstrate that RHAMM is required for the activation of Aurora kinase A. Silencing of RHAMM delays the kinetics of spindle assembly, mislocalizes targeting protein for XKlp2 (TPX2), and attenuates the localized activation of Aurora kinase A with a consequent reduction in mitotic spindle length. The RHAMM-TPX2 complex requires a C-terminal basic leucine zipper in RHAMM and a domain that includes the nuclear localization signal in TPX2. Together, our findings identify RHAMM as a critical regulator for Aurora kinase A signaling and suggest that RHAMM ensures bipolar spindle assembly and mitotic progression through the integration of biochemical and structural pathways.
Sigma-Aldrich
MISSION® esiRNA, targeting human TPX2
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tpx2
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining