Skip to Content
Merck
All Photos(1)

Key Documents

81357

Sigma-Aldrich

Poly(Glu, Tyr) sodium salt

suitable for ligand binding assays, Mol wt 20,000-50,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

Product Name

Poly(Glu, Tyr) sodium salt, mol wt 20,000-50,000, Glu:Tyr 4:1

Quality Level

form

powder

feed ratio

Glu:Tyr (4:1)

mol wt

20,000-50,000

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Poly(Glu,Tyr) (poly(Glu,Tyr) 4:1 random copolymer) is used as a substrate for the evaluation of protein tyrosine kinase(s).
Poly(Glu,Tyr) 4:1 is a polyamino acid used in the manufacture of electrochemical biosensors for detection of reactions such as Fenton induced tyrosine oxidation.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masanobu Chiku et al.
Biosensors & bioelectronics, 26(1), 235-240 (2010-07-17)
In this report, we determined protein tyrosine kinase (PTKs) activity in human epidermoid carcinoma cells (A431) by employing a novel electrochemical method using boron-doped diamond (BDD) electrodes that enables the electrochemical oxidation of tyrosine (Tyr), phosphorylated Tyr (Tyr-P) and sulfated
Morten R Clausen et al.
Journal of agricultural and food chemistry, 56(18), 8692-8698 (2008-09-02)
Oxidation of 2,2'-azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) by lactoperoxidase was found to be inhibited by tyrosine-containing random amino acid copolymers but not by tyrosine. Both electrostatic effects and polymer size were found to be important by comparison of negatively and positively charged copolymers
Na Qu et al.
Biosensors & bioelectronics, 26(5), 2292-2296 (2010-11-03)
A simple and sensitive electrochemical biosensor was used to detect tyrosine oxidation induced by hydroxyl radicals generated by Fenton reaction (Fe(2+)/H(2)O(2)). Poly(glu, tyr) (4:1) peptides were immobilized on indium tin oxide (ITO) electrode surface via layer-by-layer assembly technique, and Os(bpy)(3)(2+)-mediated
Edit Varkondi et al.
Journal of receptor and signal transduction research, 25(1), 45-56 (2005-06-18)
Receptor tyrosine kinases (PTKs) play key roles in the pathogenesis of numerous human diseases, including cancer, and therefore PTK inhibitors are currently under intense investigation as potential drug candidates. PTK inhibitor screening data are, however, poorly comparable because of the
D M Williams et al.
The Journal of biological chemistry, 275(49), 38127-38130 (2000-09-28)
Protein-tyrosine kinases contain a catalytic loop Arg residue located either two or four positions downstream of a highly conserved Asp residue. In this study, the role of this Arg (Arg-318) in the protein-tyrosine kinase C-terminal Src kinase (Csk) was investigated.

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service