Skip to Content
Merck
All Photos(1)

Key Documents

G0903

Sigma-Aldrich

γ-Glu-Cys

≥80% (HPLC), suitable for ligand binding assays

Synonym(s):

γ-L-Glutamyl-L-cysteine, des-Gly-glutathione reduced form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O5S
CAS Number:
Molecular Weight:
250.27
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

γ-Glu-Cys, ≥80% (HPLC)

Quality Level

Assay

≥80% (HPLC)

form

powder

composition

Peptide content, ≥60% elemental analysis

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CS)C(O)=O)C(O)=O

InChI

1S/C8H14N2O5S/c9-4(7(12)13)1-2-6(11)10-5(3-16)8(14)15/h4-5,16H,1-3,9H2,(H,10,11)(H,12,13)(H,14,15)/t4-,5-/m0/s1

InChI key

RITKHVBHSGLULN-WHFBIAKZSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glu-Cys

Application

γ-Glu-Cys is used to study the active sites and kinetics of glutathione synthetase(s). It has also been used for the extraction and quantification of low molecular weight thiols from Arabidopsis tissues.

Biochem/physiol Actions

γ-L-Glutamyl-L-cysteine (γ-Glu-Cys) is a substrate used for the biosynthesis of L-glutathione by glutathione synthetase(s). γ-Glu-Cys is generated by the cleavage of the Cys-Gly peptide bond of glutathione. It is also essential for the formation of phytochelatins, which are cysteine-rich thiol-reactive peptides.

Physical form

Lyophilized from 0.1% TFA.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Andersson et al.
Clinical chemistry, 38(7), 1311-1315 (1992-07-01)
The concentration of free and total homocysteine in plasma increases in time if blood is stored uncentrifuged after sampling. The increase is temperature dependent and the maximal increase in total plasma homocysteine at 37 degrees C was 3.0 mumol.L-1.h-1. Even
Quantification of Low Molecular Weight Thiols in Arabidopsis
Ziqing Miao
Bio-protocol, 6(1) (2016)
Naser A. Anjum, Sarvajeet Singh Gill, Ritu Gill
Plant Adaptation to Environmental Change: Significance of Amino Acids and their Derivatives (2014)
Chava B Pocernich et al.
Biochimica et biophysica acta, 1822(5), 625-630 (2011-10-22)
Oxidative stress has been associated with the onset and progression of mild cognitive impairment (MCI) and Alzheimer disease (AD). AD and MCI brain and plasma display extensive oxidative stress as indexed by protein oxidation, lipid peroxidation, free radical formation, DNA
Runa Njålsson et al.
Human genetics, 116(5), 384-389 (2005-02-18)
Glutathione synthetase (GS) deficiency is a rare autosomal recessive disorder. The clinical phenotype varies widely, and nearly 30 different mutations in the GSS gene have been identified. In the present study, genotype, enzyme activity, metabolite levels and clinical phenotype were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service