Skip to Content
Merck
All Photos(1)

Key Documents

76436

Sigma-Aldrich

Pentaethylene glycol monodecyl ether

BioXtra, ≥97.0% (GC)

Synonym(s):

C10E5, Decyl pentaethylene glycol ether, Decylpentaglycol, Polyoxyethylene (5) decyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)5OH
CAS Number:
Molecular Weight:
378.54
Beilstein:
2457059
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

Assay

≥97.0% (GC)

form

liquid

mol wt

378.54 g/mol

refractive index

n20/D 1.453

density

0.973 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCOCCOCCOCCOCCOCCO

InChI

1S/C20H42O6/c1-2-3-4-5-6-7-8-9-11-22-13-15-24-17-19-26-20-18-25-16-14-23-12-10-21/h21H,2-20H2,1H3

InChI key

QAXPOSPGRHYIHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Effect of Hydrostatic Pressure on Gas Solubilization in Micelles:</strong> This research investigates how hydrostatic pressure affects gas solubilization in micelles, hinting at the environmental and industrial relevance of Pentaethylene glycol monodecyl ether in creating more efficient surfactant systems (Meng and Ashbaugh, 2015).</li>
<li><strong>Structural Evolution in the Isotropic Channel of a Water-Nonionic Surfactant System That Has a Disconnected Lamellar Phase:</strong> The paper details the behavior of nonionic surfactant systems under varied conditions, useful in understanding the solubilizing and stabilizing properties of Pentaethylene glycol monodecyl ether in pharmaceutical and cosmetic products (Stubenrauch et al., 2012).</li>
</ul>

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marvin V Dilworth et al.
Methods (San Diego, Calif.), 147, 3-39 (2018-04-16)
Despite many high-profile successes, recombinant membrane protein production remains a technical challenge; it is still the case that many fewer membrane protein structures have been published than those of soluble proteins. However, progress is being made because empirical methods have
Mireia Subinya et al.
Colloids and surfaces. B, Biointerfaces, 131, 108-114 (2015-05-15)
The paper at hand deals with the influence of the pH-value on the conformation and activity of the lipase B from Candida antarctica (CalB) which is incorporated in a bicontinuous microemulsion. The microemulsion used for this purpose consists of water/NaCl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service