Skip to Content
Merck
All Photos(1)

Key Documents

75759

Sigma-Aldrich

Quercetin 3-O-α-L-arabinopyranoside

≥95% (HPLC)

Synonym(s):

3′,4′,5,7-Tetrahydroxyflavone 3-O-α-L-arabinoside, 3-(α-L-Arabinopyranosyloxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Feniculin, Foeniculin, Guaijaverin, Quercetin 3-O-α-L-arabinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

storage temp.

room temp

SMILES string

O[C@H]1CO[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H]1O

InChI

1S/C20H18O11/c21-8-4-11(24)14-13(5-8)30-18(7-1-2-9(22)10(23)3-7)19(16(14)27)31-20-17(28)15(26)12(25)6-29-20/h1-5,12,15,17,20-26,28H,6H2/t12-,15-,17+,20-/m0/s1

InChI key

PZZRDJXEMZMZFD-IEGSVRCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin 3-O-α-L-arabinopyranoside was isolated from the leaves of the Ruprechtia polystachya tree. [1]

Biochem/physiol Actions

Plant flavonoid which exhibits antibacterial and antioxidant activities. It shows significantly strong ABTS radical scavenging activity[2], inhibitory activity against urease of quercetin glycosides isolated from Allium cepa and Psidium guajava[3]. It is a potential antiplaque agent against Streptococcus mutans[4].
quercetin 3-O-α-L-arabinopyranoside was shown to inhibit the activity of glucose-6-phosphatase. [1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dimitrina Zheleva-Dimitrova et al.
Natural product research, 26(17), 1576-1583 (2011-11-15)
The occurrence of three known benzophenones, namely annulatophenonoside, acetylannulatophenonoside and annulatophenone as well as a flavonol O-glycoside guajaverin in the aerial parts of Hypericum maculatum Crantz was established. In addition, hyperoside, isoquercitrin and miquelianin were isolated from this plant, as
X Lozoya et al.
Archives of medical research, 25(1), 11-15 (1994-01-01)
The traditional herbal remedy from Psidium guajava leaves has been medically proposed in Mexico as effective treatment of acute diarrhea. A methanolic leaf extract was subjected to a bioassay-guided isolation of spasmolytic constituents. Six fractions were separated on a polyvinylpolypyrrolidine
Cássia Regina Primila Cardoso et al.
Toxicology, 225(1), 55-63 (2006-06-20)
Byrsonima crassa is a plant pertaining to the Brazilian central savannah-like belt of vegetation and popularly used for the treatment of gastric dysfunctions and diarrhoea. The methanol extract contains catechin, tannins, terpenes and flavonoids; both mutagenic potential and antioxidant properties
Maryan Bruzual De Abreu et al.
Chemistry & biodiversity, 8(11), 2126-2134 (2011-11-16)
Two new compounds, 5-methyl-2-(2-methylbutanoyl)phloroglucinol 1-O-(6-O-β-D-apiofuranosyl)-β-D-glucopyranoside (1) and trans-2,3-dihydrokaempferol 3-O-(4-O-sulfo)-α-L-arabinopyranoside (2), together with 14 known flavonoids, trans-dihydrokaempferol 3-O-α-L-arabinopyranoside (3), trans-taxifolin 3-O-α-L-arabinofuranoside (4), quercetin 3-O-α-L-rhamnopyranoside (5), quercetin 3'-O-α-L-arabinofuranoside (6), catechin 3-O-α-L-rhamnopyranoside (7), trans-taxifolin 3-O-α-L-arabinopyranoside (8), cis-dihydrokaempferol 3-O-α-L-arabinopyranoside (9), catechin (10), myricetin 3-O-α-L-rhamnopyranoside
G R Prabu et al.
Journal of applied microbiology, 101(2), 487-495 (2006-08-03)
The aim of the present study was to investigate the anti-Streptococcus mutans activity and the in vitro effects of subminimal inhibitory concentrations of guaijaverin isolated from Psidium guajava Linn. on cariogenic properties of Strep. mutans. Bioautography-directed chromatographic fractionation, yield biologically

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service