46935
6-[Fluorescein-5(6)-carboxamido]hexanoic acid
suitable for fluorescence, ≥90% (HPCE)
Synonym(s):
Fluorescein-5(6)-carboxamidocaproic acid
About This Item
Recommended Products
Assay
≥90% (HPCE)
form
solid
solubility
DMF: soluble
DMSO: soluble
fluorescence
λex 491 nm; λem 515 nm in 0.1 M Tris pH 9.0
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
OC(=O)CCCCCNC(=O)c1ccc2c(c1)C(=O)OC23c4ccc(O)cc4Oc5cc(O)ccc35.OC(=O)CCCCCNC(=O)c6ccc7C(=O)OC8(c9ccc(O)cc9Oc%10cc(O)ccc8%10)c7c6
InChI
1S/2C27H23NO8/c29-16-6-9-20-22(13-16)35-23-14-17(30)7-10-21(23)27(20)19-8-5-15(12-18(19)26(34)36-27)25(33)28-11-3-1-2-4-24(31)32;29-16-6-9-19-22(13-16)35-23-14-17(30)7-10-20(23)27(19)21-12-15(5-8-18(21)26(34)36-27)25(33)28-11-3-1-2-4-24(31)32/h2*5-10,12-14,29-30H,1-4,11H2,(H,28,33)(H,31,32)
InChI key
WEJBWDKWIFHWDE-UHFFFAOYSA-N
Application
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service