Skip to Content
Merck
All Photos(1)

Key Documents

46051

Sigma-Aldrich

Esterase from Bacillus stearothermophilus

≥0.2 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized

specific activity

≥0.2 U/mg

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

The compound is commonly used for the synthesis of biodiesel and biopolymers, as well as in the production of pharmaceuticals, agrochemicals and flavor compounds.

Biochem/physiol Actions

Esterase acts on water-soluble carboxyl esters containing short chain fatty acids .
The esterase catalyzes the transesterification of 1-phenylethanol.

Unit Definition

1 U corresponds to the amount of enzyme which releases 1 μmol 4-nitrophenol per minute at pH 7.0 and 65°C (4-nitrophenyl-n-octanoate as substrate)

Other Notes

Heat stable enzyme

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.A. Cowan
Enzyme and Microbial Technology, 12, 374-374 (1990)
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Joshua P Lewis et al.
Pharmacogenetics and genomics, 23(1), 1-8 (2012-11-01)
Carboxylesterase 1 (CES1) is the primary enzyme responsible for converting clopidogrel into biologically inactive carboxylic acid metabolites. We genotyped a functional variant in CES1, G143E, in participants of the Pharmacogenomics of Anti-Platelet Intervention (PAPI) study (n=566) and in 350 patients
Judit Marsillach et al.
Toxicology, 307, 46-54 (2012-12-25)
Exposure to organophosphorus (OP) compounds can lead to serious neurological damage or death. Following bioactivation by the liver cytochromes P450, the OP metabolites produced are potent inhibitors of serine active-site enzymes including esterases, proteases and lipases. OPs may form adducts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service