Merck
  • Comparative analysis of selected biomarkers and pesticide sensitivity in juveniles of Solea solea and Solea senegalensis.
Comparative analysis of selected biomarkers and pesticide sensitivity in juveniles of Solea solea and Solea senegalensis.
Environmental science and pollution research international (2012-12-06)
B Sànchez-Nogué, I Varó, M Solé
ABSTRACT
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison of activities on enzymes involved in xenobiotic metabolism was carried out. Juveniles of both species were sampled in winter 2011 from the Ebro Delta region, and activities of selected enzymes such as acetylcholinesterase (AChE), carboxylesterase (CbE), ethoxyresorufin O-deethylase (EROD) and glutathione S-transferase (GST) were determined in several tissues. Lipid peroxidation (LP) levels in plasma were measured as a sign of oxidative stress. In vitro exposures to selected pesticides were contrasted, analysing AChE and CbE activities in several tissue homogenates. Overall, enzymatic activities were higher in S. solea except for gill GST and CbE and kidney GST, while plasmatic LP levels were similar. In vitro contrasts revealed lower IC50 values for CbE activities in S. solea, suggesting a greater buffer capacity of this enzyme to potentially reduce pesticide toxicity over AChE.
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg