Skip to Content
Merck
All Photos(3)

Documents

3873

Sigma-Aldrich

Tartrate Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

Quality Level

500
600

IVD

for in vitro diagnostic use

concentration

335 mmol/L

pH

(4.9 +/- 0.1)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

For use in the Acid Phosphatase (Leukocyte) Kit No. 387A

Other Notes

Tartrate buffer 0.335 mol/L

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ju Pan et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 67(10), 847-861 (2018-08-16)
To investigate the amelioration effects of quetiapine on rheumatoid arthritis with RAW 264.7 macrophage and collagen-induced arthritis (CIA) DBA/1J mouse model. RAW 264.7 macrophage and DBA/1J mice. Lipopolysaccharide and collagen. RAW 264.7 macrophages stimulated by lipopolysaccharide (LPS) followed by quetiapine
John M E Nichols et al.
Developmental cell, 48(4), 491-505 (2019-01-08)
Signaling from chemoattractant receptors activates the cytoskeleton of crawling cells for chemotaxis. We show using phosphoproteomics that different chemoattractants cause phosphorylation of the same core set of around 80 proteins in Dictyostelium cells. Strikingly, the majority of these are phosphorylated
Anindya Dey et al.
Science advances, 6(27) (2020-09-17)
The stringent expression of the hypoxia inducible factor-1α (HIF-1α) is critical to a variety of pathophysiological conditions. We reveal that, in normoxia, enzymatic action of cystathionine β-synthase (CBS) produces H2S, which persulfidates prolyl hydroxylase 2 (PHD2) at residues Cys21 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service