Skip to Content
Merck
All Photos(2)

Key Documents

3863

Sigma-Aldrich

Acetate Solution

2.5 M, pH 5.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

form

solution

IVD

for in vitro diagnostic use

concentration

2.5 M

pH

5.2

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

Intended for use in the Leukocyte Acid Phosphatase procedure, procedure No. 386 or 387

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cen Wen et al.
Aging, 12(4), 3862-3879 (2020-03-07)
Perioperative neurocognitive disorders are common in elderly patients who have undergone surgical procedures. Neuroinflammation induced by microglial activation is a hallmark of these neurological disorders. Acetate can suppress inflammation in the context of inflammatory diseases. We employed an exploratory laparotomy
Mengda Xu et al.
Experimental & molecular medicine, 51(7), 83-83 (2019-07-25)
Acetate has been indicated to be elevated and to regulate inflammation in inflammatory and metabolic diseases. The inflammasome serves as a key component of immune homeostasis, and its dysregulation can lead to various inflammatory disorders. However, little is known about
Yi-Ju Pan et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 67(10), 847-861 (2018-08-16)
To investigate the amelioration effects of quetiapine on rheumatoid arthritis with RAW 264.7 macrophage and collagen-induced arthritis (CIA) DBA/1J mouse model. RAW 264.7 macrophage and DBA/1J mice. Lipopolysaccharide and collagen. RAW 264.7 macrophages stimulated by lipopolysaccharide (LPS) followed by quetiapine
Georgia Pennarossa et al.
Stem cell reviews, 14(3), 398-411 (2017-12-30)
Several studies have demonstrated the possibility to revert differentiation process, reactivating hypermethylated genes and facilitating cell transition to a different lineage. Beside the epigenetic mechanisms driving cell conversion processes, growing evidences highlight the importance of mechanical forces in supporting cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service