Skip to Content
Merck
All Photos(1)

Key Documents

357M-1

Sigma-Aldrich

TCL1 (MRQ-7) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MRQ-7, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (357M-14)
vial of 0.5 mL concentrate (357M-15)
bottle of 1.0 mL predilute (357M-17)
vial of 1.0 mL concentrate (357M-16)
bottle of 7.0 mL predilute (357M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

B-cell lymphoma, tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, nuclear

Gene Information

human ... TCL1A(8115)

General description

T-cell leukemia/lymphoma protein 1 (TCL1, TCL1A, p14TCL1) is a 14 kDa product of the TCL1 gene that is involved in T-cell prolymphocytic leukemia (T-PLL). TCL1 protein is normally found in the nucleus and cytoplasm of lymphoid lineage cells during early embryogenesis. Chromosomal translocations may lead to overexpression of TCL1, resulting in T-cell leukemia and B-cell lymphoma. TCL1 binds to a novel site in the pleckstrin homology (PH) domain, resulting in activation and nuclear translocation of Akt by overexpressed TCL1 which may promote an anti-apoptotic response; this may normally serve to promote growth during development but may lead to malignancy when TCL1 is overexpressed. TCL1 is expressed in differentiated B-cells under both reactive and neoplastic conditions, antigen committed B-cells, and in germinal center B-cells. TCL1 is down-regulated in the latest stage of B-cell differentiation. TCL1 is overexpressed in Burkitt lymphoma, the majority of AIDS-related non-Hodgkin lymphoma-designated immunoblastic plasmacytoid lymphoma, lymphoblastic lymphoma, chronic lymphocytic leukemia, mantle cell lymphoma, follicular lymphoma, diffuse large B-cell lymphoma, and primary cutaneous B-cell lymphoma. Therefore, the most useful application of anti-TCL1 is the discrimination of B-cell lymphomas from T-cell lymphomas, CD30+ anaplastic large cell lymphomas, multiple myeloma, and marginal zone B-cell lymphoma.

Quality


IVD

IVD

IVD

RUO

Linkage

TCL1 Positive Control Slides, Product No. 357S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Pescarmona et al.
Histopathology, 49(4), 343-348 (2006-09-19)
To investigate by immunohistochemical analysis the expression of the TCL1 oncogene product and of CD27 in 25 cases of primary cutaneous B-cell lymphomas (PCBCL) classified according to the World Health Organization-European Organization for Research and Treatment of Cancer classification of
M G Narducci et al.
Cancer research, 60(8), 2095-2100 (2000-04-29)
Chromosomal rearrangements observed in T-cell prolymphocytic leukemia involve the translocation of one T-cell receptor gene to either chromosome 14q32 or Xq28, deregulating the expression of cellular protooncogenes of unknown function, such as TCL1 or its homologue, MTCP1. In the human
J Roos et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 69(2), 59-66 (2001-12-26)
TCL1, MTCP1 and TCL1b are three members of a new family of oncogenes that are expressed in T cell leukemias of ataxia telangiectasia patients (T-PLL, T-CLL). TCL1 is located at 14q32.1 and activated by juxtaposition to the alpha/delta-locus at 14q11
I Nakayama et al.
Pathology international, 50(3), 191-199 (2000-05-03)
The TCL1 gene, localized near the break point of chromosome 14q32.1 often involved in T cell leukemias, is also expressed in normal precursor T and B cells, and B cell lymphoma cell lines. We investigated the expression of the TCL1
Yuri Pekarsky et al.
Hematology/oncology clinics of North America, 18(4), 863-879 (2004-08-25)
Mature leukemias and lymphomas are a heterogeneous group of lymphoproliferative neoplasias characterized by the accumulation of mature lymphocytes in lymph nodes, other lymphoid tissues, and peripheral blood. In this article we discuss molecular mechanisms leading to the pathogenesis of two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service