Skip to Content
Merck
All Photos(1)

Key Documents

321M-1

Sigma-Aldrich

PLAP (NB10) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

diluted ascites fluid

antibody product type

primary antibodies

clone

NB10, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (321M-14)
vial of 0.5 mL concentrate (321M-15)
bottle of 1.0 mL predilute (321M-17)
vial of 1.0 mL concentrate (321M-16)
bottle of 7.0 mL predilute (321M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgGκ

control

placenta

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... ALPP(250)

General description

Placental Alkaline Phosphatase (PLAP) belongs to a family of four isozymes that are synthesized in primordial germ cells and in term placental syncytiotrophoblasts. Anti-PLAP may be useful in the identification of germ cell tumors.

Linkage

PLAP Positive Control Slides, Product No. 321S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Burke et al.
Human pathology, 19(6), 663-670 (1988-06-01)
Two hundred three testicular germ cell tumors were studied immunohistochemically for the presence of placental alkaline phosphatase (PLAP). Special emphasis was placed on the pattern and incidence of positive staining of intratubular malignant germ cells (ITMGCs) adjacent to tumors. 99%
G K Jacobsen et al.
Acta pathologica, microbiologica, et immunologica Scandinavica. Section A, Pathology, 92(5), 323-329 (1984-09-01)
Recently, placental alkaline phosphatase (PLAP) has been suggested as a tumour marker in patients with seminomas (S), since elevated serum levels of PLAP were found with high frequency in these patients. The present immunoperoxidase study of 33 testicular germ cell
J D Wencel-Drake et al.
The American journal of pathology, 115(2), 156-164 (1984-05-01)
The distribution and transport of thrombospondin (TSP), fibrinogen (Fbg), fibronectin (Fn), and Factor VIII-related antigen (VIII:RAg) in resting and thrombin-stimulated platelets was investigated by immunofluorescence microscopy. In resting intact cells, little surface staining was seen for these proteins. In permeable
Jeffrey D Goldsmith et al.
The American journal of surgical pathology, 26(12), 1627-1633 (2002-12-03)
Placental alkaline phosphatase (PLAP) is normally produced by primordial germ cells and syncytiotrophoblasts, and the detection of its expression has been useful in the diagnosis of germ cell tumors. We have recently observed PLAP immunoreactivity in normal human adult and
A Lösch et al.
Acta obstetricia et gynecologica Scandinavica, 75(8), 753-756 (1996-09-01)
The gestational trophoblastic disease summarizes all types of hydatidiform moles, placental site trophoblastic tumor and choriocarcinoma. It is of clinical relevance to distinguish between complete hydatidiform mole and partial hydatidiform mole to predict prognosis of recurrency of molar pregnancy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service