Skip to Content
Merck
All Photos(1)

Key Documents

322M-1

Sigma-Aldrich

Podoplanin (D2-40) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

D2-40, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (322M-14)
vial of 0.5 mL concentrate (322M-15)
bottle of 1.0 mL predilute (322M-17)
vial of 1.0 mL concentrate (322M-16)
bottle of 7.0 mL predilute (322M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20-1:100

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... PDPN(10630)

General description

Podoplanin is a transmembrane mucoprotein (38 kD) recognized by the D2-40 monoclonal antibody. Podoplanin is selectively expressed in normal and malignant lymphatic endothelium, including Kaposi’s sarcomas. Podoplanin has also been shown to be expressed in epithelioid mesotheliomas and seminomas.

Quality


IVD

IVD

IVD

RUO

Linkage

Podoplanin Positive Control Slides, Product No. 322S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Csaba Galambos et al.
Pediatric and developmental pathology : the official journal of the Society for Pediatric Pathology and the Paediatric Pathology Society, 8(2), 181-189 (2005-02-19)
The distinction between lymphatic and other vascular vessels on microscopic sections is a challenging task. D2-40, a novel antibody, has been reported to be selective for lymphatic endothelium. We studied the specificity and sensitivity of D2-40 in pediatric vascular tumors
Nelson G Ordóñez
Human pathology, 36(4), 372-380 (2005-05-14)
Recent investigations have shown that podoplanin and the D2-40 monoclonal antibody, which reacts with an oncofetal antigen present in fetal germ cells, are highly reliable lymphatic endothelial markers. The observation that both of these markers are also expressed in normal
Nelson G Ordóñez
Advances in anatomic pathology, 13(2), 83-88 (2006-05-04)
Podoplanin is a transmembrane mucoprotein recognized by the recently commercially available D2-40 monoclonal antibody. Recent investigations have shown that podoplanin is selectively expressed in lymphatic endothelium as well as lymphangiomas, Kaposi sarcomas, and in a subset of angiosarcomas with probable
Folker E Franke et al.
Journal of cutaneous pathology, 31(5), 362-367 (2004-04-03)
Hobnail hemangioma (targetoid hemosiderotic hemangioma) is a small benign vascular tumor of the superficial and mid-dermis. In contrast to its well-characterized histology, it has been unclear whether this tumor arises from blood vessel endothelial cells (BECs) or lymphatic vessel endothelial
F Fogt et al.
Oncology reports, 11(1), 47-50 (2003-12-05)
Distribution of lymphatic vessels in normal and neoplastic colon has been previously analyzed with electron microscopic techniques, as reliable antibodies have not been available for selective lymph vessel staining. A novel monoclonal antibody, D2-40, is recently available to differentiate lymphatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service