Skip to Content
Merck
All Photos(1)

Key Documents

31995

Sigma-Aldrich

Barbital

BioXtra, ≥99.0% (T)

Synonym(s):

5,5-Diethyl-2,4,6(1H,3H,5H)-pyrimidinetrione, 5,5-Diethylbarbituric acid, Barbitone, Veronal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2O3
CAS Number:
Molecular Weight:
184.19
Beilstein:
163999
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21

product line

BioXtra

Quality Level

Assay

≥99.0% (T)

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 20 °C (HV)

mp

189-191 °C

solubility

alcohol: soluble 1 gm in 14 ml
chloroform: soluble 1 gm in 75 ml
diethyl ether: soluble 1 gm in 35 ml
water: soluble 1 gm in 13 ml (boiling)
water: soluble 1 gm in 130 ml
acetone: soluble
ethyl acetate: soluble
nitrobenzene: soluble
petroleum ether: soluble
pyridine: soluble

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

SMILES string

CCC1(CC)C(=O)NC(=O)NC1=O

InChI

1S/C8H12N2O3/c1-3-8(4-2)5(11)9-7(13)10-6(8)12/h3-4H2,1-2H3,(H2,9,10,11,12,13)

InChI key

FTOAOBMCPZCFFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Barbital has been used as an assay buffer for determining the carbonic anhydrase activity. It has also been used for rocket immunoelectrophoresis.

Biochem/physiol Actions

Barbital also referred to as 5,5-Diethylbarbituric acid, Barbitone or Veronal is a sedative/hypnotic (sleeping aid). It stimulates the development of tumors in rats treated with a sub-effective dose of N-bis(2-hydroxypropyl)nitrosamine (DHPN).
Sedative/hypnotic

Other Notes

Sales restrictions may apply

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crab Gill Intra-Epithelial Carbonic Anhydrase Plays a Major Role in Haemolymph CO2 and Chloride Ion Regulation.
Louis E. Burnett
The Journal of Experimental Biology, 92, 243-254 (1981)
Assay for Cauliflower Mosaic Virus.
Hagen, T. J
Phytopathology, 72, 239-242 (1982)
Y Hiasa et al.
Carcinogenesis, 3(10), 1187-1190 (1982-01-01)
Phenobarbital (PB) and barbital (BB) promoted the development of thyroid tumors in rats treated with a sub-effective dose of N-bis(2-hydroxypropyl)nitrosamine (DHPN) for thyroid tumorigenesis. Rats were given s.c. injections of 70 mg DHPN/100 g body weight once a week for
J M Bossert et al.
Behavioural pharmacology, 14(7), 517-523 (2003-10-15)
We have shown previously that 15 mg/kg pentobarbital induces a conditioned place preference (CPP), but it is unsuitable for intracranial administration. Since the long-acting barbiturate, sodium barbital, is soluble at a neutral pH, we tested whether it would induce a
Marlies Oostendorp et al.
Transfusion, 55(6 Pt 2), 1555-1562 (2015-05-20)
Monoclonal antibodies (MoAbs) are increasingly integrated in the standard of care. The notion that therapeutic MoAbs can interfere with clinical laboratory tests is an emerging concern that requires immediate recognition and the development of appropriate solutions. Here, we describe that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service