Skip to Content
Merck
All Photos(1)

Key Documents

11715

Sigma-Aldrich

Sodium barbiturate

≥97.0% (T)

Synonym(s):

Barbituric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N2NaO3
CAS Number:
Molecular Weight:
150.07
Beilstein:
3748030
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

solubility

hot water: soluble 0.2 g/10 mL, clear, colorless

SMILES string

[Na+].[O-]C1=NC(=O)NC(=O)C1

InChI

1S/C4H4N2O3.Na/c7-2-1-3(8)6-4(9)5-2;/h1H2,(H2,5,6,7,8,9);/q;+1/p-1

InChI key

MHQHHBYRYFICDV-UHFFFAOYSA-M

General description

Sodium barbiturate is a white to light beige powder. It accelerates the propagation of metastatic prostate adenocarcinomas in rats and in tissue culture.

Application

Sodium barbiturate has been used for quantification of thrombin in plasma. It has been used in the estimation of trypsin in duodenal contents. It was also used as a buffer to characterise the functions of two long-chain fatty acid CoA ligase genes (facl) in crude oil-degrading Geobacillus thermodenitrificans NG80-2.

Other Notes

Sales restrictions may apply

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

F A Ofosu et al.
Blood, 64(3), 742-747 (1984-09-01)
Heparan with a low affinity for antithrombin III has previously been demonstrated to inhibit thrombin generation in both normal plasma and plasma depleted of antithrombin III. In addition, standard heparin and heparin with a low affinity for antithrombin III have
M Pollard et al.
Oncology, 34(3), 129-132 (1977-01-01)
Metastatic prostate adenocarcinomas, derived from aging germfree Wistar rats, have been propagated in rats and in tissue culture. A protocol has been developed and demonstrated for assay of treatments which retard or which accelerate the rate and extent of tumor
Simple method for estimating trypsin.
H S Wiggins
Gut, 8(4), 415-416 (1967-08-01)
Thomas Gelbrich et al.
Acta crystallographica. Section C, Crystal structure communications, 66(Pt 1), o55-o58 (2010-01-06)
Both title structures exhibit essentially planar barbiturate rings. The crystal structure of enallylpropymal (5-allyl-5-isopropyl-1-methylbarbituric acid), C(11)H(16)N(2)O(3), is composed of centrosymmetric N-H...O hydrogen-bonded dimers, while 1,5-di(but-2-enyl)-5-ethylbarbituric acid, C(14)H(20)N(2)O(3), forms N-H...O hydrogen-bonded helical chains. Each of the ten known crystal structures of
Peter Ballard et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(12), 2165-2168 (2011-09-02)
Accurately predicting in vivo metabolic clearance from in vitro liver microsomes or hepatocytes requires a good understanding of the factors contributing to the prediction. Although much work has concentrated on deriving scaling factors and optimizing the metabolic stability techniques for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service