Skip to Content
Merck
All Photos(3)

Key Documents

227R-2

Sigma-Aldrich

BCL6 (EP278) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP278, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (227R-27)
bottle of 7.0 mL predilute (227R-28)
vial of 0.1 mL concentrate (227R-24)
vial of 0.5 mL concentrate (227R-25)
vial of 1.0 mL concentrate (227R-26)
vial of 0.1 mL concentrate Research Use Only (227R-24-RUO)
vial of 1.0 mL concentrate Research Use Only (227R-26-RUO)
vial of 1.0 mL pre-dilute Research Use Only (227R-27-RUO)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (227R-28-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... BCL6(604)

General description

BCL6 is a transcriptional regulator gene which codes for a 706-amino-acid nuclear zinc finger protein.1 In normal tissue these antibodies have strong nuclear staining for a subset of B-lymphocytes, mostly located in germinal centers (GC).2 BCL6 antibodies stain malignant cells in follicular lymphoma,3 diffuse large B-cell lymphomas,4 Burkitt lymphoma,4 classical Hodgkin lymphoma,5 as well as majority of tumor cells in nodular lymphocyte predominant Hodgkin lymphoma.6 BCL6 expression has been also seen in anaplastic large cell lymphomas (ALCL).7

Quality


IVD

IVD

IVD

RUO

Linkage

BCL6 Positive Control Slides, Product No. 227S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bai et al.
Leukemia & lymphoma, 47(3), 495-501 (2006-01-07)
The bcl6/CD10/MUM1/CD138 B-cell differentiation immunophenotypes were analysed in 101 cases of classical Hodgkin lymphomas (cHL) aiming to elucidate their histogenesis. Three major bcl6/CD10/MUM1/CD138 immunophenotypes were distinguished on the basis of the immunohistochemical positivity of Hodgkin and Reed-Sternberg (H/RS) cells: (a)
Arzu Saglam et al.
Tumori, 97(5), 634-638 (2011-12-14)
Several transcription factors predominantly used for B-cell lineage identification are also expressed in a small percentage of T cells within germinal centers and interfollicular areas. The aim of the study was to evaluate the expression of Mum-1, Oct-2 and Bcl-6
Expression of BCL-6 protein and CD138/syndecan-1 as B-cell markers in Hodgkin's disease.
A Carbone et al.
The International journal of biological markers, 14(3), 144-148 (1999-11-24)
A Carbone et al.
Blood, 90(6), 2445-2450 (1997-10-06)
The expression pattern of the BCL-6 transcription factor has been assessed in normal and neoplastic B-cell populations and in Hodgkin's disease. However, little is known about BCL-6 expression and its biological significance in T-cell neoplasms. In this study, a series
A Carbone et al.
Blood, 91(3), 747-755 (1998-02-03)
This study was aimed at defining the histogenesis of the pathologic spectrum of acquired immunodeficiency syndrome-related non-Hodgkin's lymphomas (AIDS-NHL), including AIDS-related small noncleaved cell lymphoma (AIDS-SNCCL), AIDS-related large noncleaved cell lymphoma (AIDS-LNCCL), AIDS-related large cell immunoblastic lymphoma plasmacytoid (AIDS-IBLP), and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service