Skip to Content
Merck
All Photos(1)

Key Documents

22075

Sigma-Aldrich

(−)-trans-Caryophyllene

(−)-trans-Caryophyllene

≥98.0% (sum of enantiomers, GC)

Synonym(s):

β-Caryophyllene, trans-(1R,9S)-8-Methylene-4,11,11-trimethylbicyclo[7.2.0]undec-4-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2044564
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −10±1°, neat

refractive index

n20/D 1.499

bp

262-264 °C (lit.)

density

0.902 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@@]12CC\C(C)=C\CCC(=C)[C@H]1CC2(C)C

InChI

1S/C15H24/c1-11-6-5-7-12(2)13-10-15(3,4)14(13)9-8-11/h6,13-14H,2,5,7-10H2,1,3-4H3/b11-6+/t13-,14-/m1/s1

InChI key

NPNUFJAVOOONJE-GFUGXAQUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-trans-Caryophyllene, a sesquiterpene, is the major constituent in the essential oil of Mentha longifolia[1] and Commiphora gileadensis.[2] It shows anti-inflammatory, local anaesthetic and antifungal properties. (-)-trans-Caryophyllene also shows potent cytotoxic activity.[2]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.9 °F - closed cup

Flash Point(C)

105.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seasonal Variation in Content and Chemical Composition of Essential Oils from Leaves of Mentha longifolia Huds.(Lamiaceae).
Niksic H, et al.
Bulletin of the Chemists and Technologists of Bosnia and Herzegovina, 2014, 29-34 null
?-Caryophyllene, a compound isolated from the biblical balm of gilead (Commiphora gileadensis), is a selective apoptosis inducer for tumor cell lines.
Amiel E, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2012 (2012)
Fabienne Hilgers et al.
Microorganisms, 9(1) (2021-01-21)
Terpenoids constitute one of the largest and most diverse groups within the class of secondary metabolites, comprising over 80,000 compounds. They not only exhibit important functions in plant physiology but also have commercial potential in the biotechnological, pharmaceutical, and agricultural
Linus Svenberg et al.
Plants (Basel, Switzerland), 10(11) (2021-11-28)
In recent work, it was shown that the graminoid plants Cynodon dactylon (Poaceae), Cyperus exaltatus (Cyperaceae), and Panicum repens (Poaceae) have an ovipositional effect on the malaria vector Anopheles gambiae in olfactometric bioassays. In order to get a view of
Getachew E Bokore et al.
Parasites & vectors, 14(1), 552-552 (2021-10-29)
Understanding the ecology and behaviour of disease vectors, including the olfactory cues used to orient and select hosts and egg-laying sites, are essential for the development of novel, insecticide-free control tools. Selected graminoid plants have been shown to release volatile

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service