Skip to Content
Merck
All Photos(1)

Key Documents

17470

Sigma-Aldrich

Bromocresol Green Sultone Form

for microscopy (Bot., Hist., Vit.), indicator (pH 3.8-5.4)

Synonym(s):

3′,3′′,5′,5′′-Tetrabromo-m-cresolsulfonephthalein, Bromocresol Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14Br4O5S
CAS Number:
Molecular Weight:
698.01
Beilstein:
372527
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bot., Hist., Vit.)
indicator (pH 3.8-5.4)

Quality Level

form

powder

technique(s)

titration: suitable

pH

3.8-5.4, yellow to blue

mp

225 °C

solubility

methanol: water (1:1): 0.1 g/10 mL, red

εmax

≥225 at 418-428 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1c(Br)c(O)c(Br)cc1C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4C

InChI

1S/C21H14Br4O5S/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21/h3-8,26-27H,1-2H3

InChI key

FRPHFZCDPYBUAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for complexometry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yu-Chang Chang et al.
Sensors (Basel, Switzerland), 11(4), 4060-4072 (2011-12-14)
A meso-structured Al-MCM-41 material was impregnated with bromocresol green (BG) dye and then incorporated into a UV-Vis DRA spectroscopic instrument for the online detection of ammonia gas. The absorption response of the Al-MCM-41/BG ammonia sensing material was very sensitive at
Kohji Hirata et al.
Research communications in molecular pathology and pharmacology, 117-118, 115-124 (2008-04-23)
Administration of antitumor drugs such as 5-fluorouracil and methotrexate to rats causes intestinal injury. The intestinal injury was reported to be prevented by coadministration of prostaglandin E1 analogues to rats. IEC-6 cell is a useful in vitro model to study
Kerstin Müller et al.
Veterinary clinical pathology, 39(1), 79-82 (2009-08-26)
In reptile medicine, plasma chemistry analysis is widely used for the evaluation of an individual's health status. The standard method for the determination of plasma albumin concentration is protein electrophoresis combined with the determination of total protein concentration, but the
Carolyn Cray et al.
Journal of avian medicine and surgery, 25(1), 14-22 (2011-06-11)
The purpose of this study was to examine the validity of albumin determinations in penguin plasma by the bromcresol green (BCG) method and the gold standard of protein electrophoresis (EPH). Plasma from 96 clinically normal and abnormal penguins (Spheniscus species)
Kai Yu et al.
Enzyme and microbial technology, 49(3), 272-276 (2011-11-25)
A pH-sensitive colorimetric assay has been established to quantitatively measure glutamate decarboxylase (GAD) activity in bacterial cell extracts using a microplate format. GAD catalyzes the irreversible α-decarboxylation of L-glutamate to γ-aminobutyrate. The assay is based on the color change of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service