Skip to Content
Merck
All Photos(1)

Key Documents

16733

Sigma-Aldrich

Quercetin 3-O-(6″-O-malonyl)-β-D-glucoside

≥85% (HPLC)

Synonym(s):

Quercetin 3-O-malonylglucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22O15
CAS Number:
Molecular Weight:
550.42
Beilstein:
6625918
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥85% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](O)[C@@H](COC(=O)CC(O)=O)O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@@H]1O

InChI

1S/C24H22O15/c25-9-4-12(28)17-13(5-9)37-22(8-1-2-10(26)11(27)3-8)23(19(17)33)39-24-21(35)20(34)18(32)14(38-24)7-36-16(31)6-15(29)30/h1-5,14,18,20-21,24-28,32,34-35H,6-7H2,(H,29,30)/t14-,18-,20+,21-,24+/m1/s1

InChI key

NBQPHANHNTWDML-UJKBSQBPSA-N

General description

The flavonoid quercetin 3-O-(6-O-malonyl)glucoside, a conjugate of quercetin, was identified in the green-leafed lettuce Lactuca sativum. [1]

Biochem/physiol Actions

Metabolite in flavone and flavonol biosynthesis, is found in plants like the fruit peel of Sicana odorifera and mulberry leaves, shows antioxidant activity.[2] Q3MG also potentially has antiatherogenic protective effects[3]; and found to improve hyperglycemia in obese mice and reduced oxidative stress in the liver.[4]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S DuPont et al.
Journal of agricultural and food chemistry, 48(9), 3957-3964 (2000-09-20)
Eight varieties of lettuce (Lactuca sativum) and three varieties of endive (Cichorium endivia) were analyzed for flavonoid composition and content. Total flavonoid contents, expressed as units of aglycon for fresh material, were in the ranges of 0.3-229 microg/g for lettuce
Byambaa Enkhmaa et al.
The Journal of nutrition, 135(4), 729-734 (2005-03-30)
The effects of dietary consumption of mulberry (Morus alba L.) leaves and their major flavonol glycoside, quercetin 3-(6-malonylglucoside) (Q3MG), on the development of atherosclerotic lesions, in relation to the susceptibility of plasma LDL to oxidative modification, was studied in LDL
Katherine Jaramillo et al.
Journal of agricultural and food chemistry, 59(3), 975-983 (2011-01-20)
Ten flavonols and three anthocyanins were identified in the fruit peel of melón de olor (Sicana odorifera), and their structures were established by spectrometric and spectroscopic (ESI-MS and NMR) techniques. One of the identified flavonols, quercetin 3-O-(6''-O-malonyl)-β-D-glucopyranoside 4'-O-β-D-glucopyranoside, has not
Takuya Katsube et al.
Journal of the science of food and agriculture, 90(14), 2386-2392 (2010-07-22)
Mulberry therapies on type 2 diabetic patients or streptozotocin-induced diabetic rats have been reported to improve fasting blood glucose levels. We investigated the effects of dietary consumption of mulberry-leaf powder and purified quercetin 3-(6-malonylglucoside), the quantitatively major flavonol glycoside in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service