Skip to Content
Merck
All Photos(1)

Key Documents

C9008

Sigma-Aldrich

p-Coumaric acid

≥98% purity (HPLC), powder

Synonym(s):

trans-4-Hydroxycinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Beilstein:
2207383
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

p-Coumaric acid, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

technique(s)

titration: suitable

color

white to yellow

mp

214 °C (dec.) (lit.)

solubility

ethanol: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)\C=C\c1ccc(O)cc1

InChI

1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+

InChI key

NGSWKAQJJWESNS-ZZXKWVIFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

p-Coumaric acid has been used as a component of chemiluminescent substrate for protein detection in western blotting.

Biochem/physiol Actions

Hydroxycinnamic acid found in many fruits and vegetables.
In western blot, p-coumaric is also used as an enhancer in chemiluminescent substrate.

Other Notes

Predominantly trans isomer.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammation triggers high mobility group box 1 (HMGB1) secretion in adipose tissue, a potential link to obesity.
Gunasekaran MK, et al.
Cytokine, 64, 103-111 (2013)
A cost effective non-commercial ECL-solution for Western blot detections yielding strong signals and low background.
Haan C and Behrmann I
Journal of Immunological Methods, 318, 11-11 (2007)
p-Coumaric acid kills bacteria through dual damage mechanisms.
Zaixiang L, et al.
Food Control, 25, 550-554 (2012)
TGF-beta receptor levels regulate the specificity of signaling pathway activation and biological effects of TGF-beta.
Rojas A, et al.
Biochimica et Biophysica Acta, 1793, 1165-1173 (2009)
p-Coumaric acid, a common dietary phenol, inhibits platelet activity in vitro and in vivo.
Luceri C, et al.
The British Journal of Nutrition, 97, 458-463 (2007)

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service