Skip to Content
Merck
All Photos(1)

Key Documents

04529

Sigma-Aldrich

Nitrilase

recombinant, expressed in E. coli, ≥2.0 U/mg

Synonym(s):

Nitrile aminohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

crystals
powder

specific activity

≥2.0 U/mg

mol wt

41 kDa

storage temp.

2-8°C

General description

Nitrilase belongs to the C-N hydrolase superfamily. It possesses the catalytic triad motifs: glutamate, lysine, and cysteine and are distributed in fungi, plants, and animals.The isoelectric point (pI) of this protein is 8.1. Native polyacrylamide gel electrophoresis (PAGE).

Application

Nitrilase may be used for immobilization on acryloyl crosslinked cellulose dialdehyde (ACCD) for catalytic activity studies.

Biochem/physiol Actions

Nitrilases catalyze the liberation of ammonia and the formation of carboxylic acid from the nitriles. The oligomeric structural organization of nitrilase is crucial for its functionality. It displays broad substrate specificity.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol ammonia per minute at pH 7.2 and 25°C with the conversion of acrylonitrile to acrylic acid

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Qiang Liu et al.
PloS one, 8(6), e67197-e67197 (2013-07-05)
In this study, several nitrilase genes from phylogenetically distinct organisms were expressed and purified in E. coli in order to study their ability to mediate the biotransformation of nitriles. We identified three nitrilases: Acidovorax facilis nitrilase (AcN); Alcaligenes fecalis nitrilase
Pablo Soriano-Maldonado et al.
Applied and environmental microbiology, 77(16), 5761-5769 (2011-06-28)
Formamidases (EC 3.5.1.49) are poorly characterized proteins. In spite of this scarce knowledge, ammonia has been described as playing a central role in the pathogenesis of human pathogens such as Helicobacter pylori, for which formamidase has been shown to participate
Y V D Nageshwar et al.
Bioprocess and biosystems engineering, 34(5), 515-523 (2010-12-29)
Microbial nitrilases are biocatalysts of interest and the enzyme produced using various inducers exhibits altered substrate specificity, which is of great interest in bioprocess development. The aim of the present study is to investigate the nitrilase-producing Alcaligenes faecalis MTCC 10757
Zhi-Jun Zhang et al.
Journal of biotechnology, 152(1-2), 24-29 (2011-01-26)
The enantioselective hydrolysis of mandelonitrile with whole cells of a recombinant Escherichia coli expressing nitrilase activity was severely inhibited by the substrate at high concentrations (>300mM), which resulted in a low yield of the target product (R)-(-)-mandelic acid. To relieve
Chunsheng Yang et al.
Applied biochemistry and biotechnology, 165(7-8), 1556-1567 (2011-09-23)
The characteristics of the new nitrilase-producing strain Rhodobacter sphaeroides LHS-305 were investigated. By investigating several parameters influencing nitrilase production, the specific cell activity was ultimately increased from 24.5 to 75.0 μmol g(-1) min(-1), and hereinto, the choice of inducer proved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service