Skip to Content
Merck
All Photos(1)

Key Documents

40972

Sigma-Aldrich

ω-Transaminase, Aspergillus terreus

recombinant, expressed in E. coli, ≥0.10 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

powder

specific activity

≥0.10 U/mg

storage temp.

−20°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which releases 1 μmol acetophenone per minute at 30°C. (R(−)-α-methyl-benzylamine as substrate).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

omega-Amino acid-pyruvate aminotransferase.
K Yonaha et al.
Methods in enzymology, 143, 500-504 (1987-01-01)
K Yonaha et al.
The Journal of biological chemistry, 267(18), 12506-12510 (1992-06-25)
The complete amino acid sequence of bacterial omega-amino acid:pyruvate aminotransferase (omega-APT) was determined from its primary structure. The enzyme protein was fragmented by CNBr cleavage, trypsin, and Staphylococcus aureus V8 digestions. The peptides were purified and sequenced by Edman degradation.
J-S Shin et al.
Applied microbiology and biotechnology, 61(5-6), 463-471 (2003-04-11)
A transaminase from Vibrio fluvialis JS17 showing activity toward chiral amines was purified to homogeneity and its enzymatic properties were characterized. The transaminase showed an apparent molecular mass of 100 kDa as determined by gel filtration chromatography and a subunit
J S Shin et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1782-1788 (2001-10-02)
Microorganisms that are capable of (S)-enantioselective transamination of chiral amines were isolated from soil samples by selective enrichment using (S)-alpha-methyl-benzylamine ((S)-alpha-MBA) as a sole nitrogen source. Among them, Klebsiella pneumoniae JS2F, Bacillus thuringiensis JS64, and Vibrio fluvialis JS17 showed good
N Watanabe et al.
Journal of biochemistry, 105(1), 1-3 (1989-01-01)
The three-dimensional structure of omega-amino acid:pyruvate aminotransferase from Pseudomonas sp. F-126, an isologous alpha 4 tetramer containing pyridoxal 5'-phosphate (PLP) as a cofactor, has been determined at 2.0 A resolution. The diffraction data were collected with a newly developed Weissenberg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service