Skip to Content
Merck
All Photos(3)

Key Documents

P7754

Sigma-Aldrich

Acetylacetone

ReagentPlus®, ≥99%

Synonym(s):

2,4-Pentanedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
741937
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021208
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
140.4 °C (lit.)
vapor pressure:
6 mmHg ( 20 °C)

grade

reagent

Quality Level

vapor density

3.5 (vs air)

vapor pressure

6 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

662 °F

expl. lim.

11.4 %

dilution

(for general lab use)

refractive index

n20/D 1.452 (lit.)

pH

6 (20 °C, 200 g/L)

bp

140.4 °C (lit.)

mp

−23 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(C)=O

InChI

1S/C5H8O2/c1-4(6)3-5(2)7/h3H2,1-2H3

InChI key

YRKCREAYFQTBPV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Acetylacetone (2,4-pentanedione) is an organic compound containing two carbonyl groups and one active methylene group. It is mainly used as an intermediate in the synthesis of various chemical derivatives. It can also be used as a modifier for polyolefins, corrosion inhibitors, and labeling of radiotracers.

Application

Acetylacetone can be used as:
  • A multifunctional ligand in the synthesis and feasible functionalization of gold nanoparticles (AuNPs).
  • A reactant to synthesize 9,10-dihydroacridines by reacting with methyl acetoacetate and Morita-Baylis-Hillman acetates.
  • A reagent in the synthesis of ZrO2(zirconium dioxide) via hydrolysis of Zr(OC3H7n)4. Acetylacetone controls the hydrolysis and condensation rates of alkoxides and thus, the nucleation and growth rates of oxides.

Packaging

Packaged in glass bottles

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rolando R Lozada-García et al.
Physical chemistry chemical physics : PCCP, 14(10), 3450-3459 (2012-02-07)
The photochemistry of the chelated enol form of acetylacetone (AcAc) was investigated by UV excitation of the S(2) state at 266 nm in parahydrogen matrices, complemented by experiments in neon and normal hydrogen matrices. Infrared (IR) spectroscopy, combined with theoretical
Heaweon Park et al.
Inorganic chemistry, 50(23), 11978-11989 (2011-11-01)
A series of high-spin iron(II) β-diketonato complexes have been prepared and characterized with the intent of modeling the substrate-bound form of the enzyme acetylacetone dioxygenase (Dke1). The Dke1 active site features an Fe(II) center coordinated by three histidine residues in
Zhenkun Sun et al.
Journal of the American Chemical Society, 134(42), 17653-17660 (2012-10-02)
The organization of different nano objects with tunable sizes, morphologies, and functions into integrated nanostructures is critical to the development of novel nanosystems that display high performances in sensing, catalysis, and so on. Herein, using acetylacetone as a chelating agent
Senthil Narayanaperumal et al.
Ultrasonics sonochemistry, 20(3), 793-798 (2012-12-12)
A task-specific ionic liquid (TSIL) has been introduced as a recyclable catalyst in Michael addition. A series of nitroalkenes and various C-based nucleophiles were reacted in the presence of 30mol% of recyclable basic-functionalized ionic liquid. Good to excellent yields were
Ivelina Georgieva et al.
Journal of molecular modeling, 18(6), 2409-2422 (2011-10-13)
Theoretical and spectroscopic studies of a series of monomeric and dimeric complexes formed through the modification of a zirconium butoxide precursor with acetylacetone and subsequent hydrolysis and/or condensation have been performed by applying DFT/B3LYP/6-31++G(d) and highly accurate RI-ADC(2) methods as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service