Skip to Content
Merck
All Photos(2)

Key Documents

P2588

Sigma-Aldrich

Potassium dichromate

BioXtra, ≥99.5%

Synonym(s):

Potassium bichromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder or crystals
solubility:
H2O: 0.1 M at 20 °C, clear, orange

grade

for analytical purposes

product line

BioXtra

Assay

≥99.5%

form

powder or crystals

reaction suitability

reagent type: oxidant

impurities

<0.005% Phosphorus (P)
<0.1% Insoluble matter

pH

3.6 (100 g/L)

mp

398 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, orange

anion traces

sulfate (SO42-): <0.005%

cation traces

Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.001%
Mg: <0.0005%
Na: <0.05%
Pb: <0.001%
Zn: ≤0.001%

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium dichromate, a toxic hexavalent chromium compound, is a strong oxidizing agent. Its nephrotoxicity in humans and animals has been studied. Its crystalline structure has been analyzed. Its ability to induce genetic and DNA methylation alteration has been assessed in Brassica napus L. plants.

Application

Potassium dichromate has been used in experiments to determine the ability of MCM-41-supported nanoscale zero-valent iron (nZVI) sample to remove hexavalent chromium.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Labra et al.
Chemosphere, 54(8), 1049-1058 (2003-12-11)
This study evaluated genetic and DNA methylation alteration induced by potassium dichromate in Brassica napus L. plants. Amplified fragment length polymorphism (AFLP) and selective amplification of polymorphic loci (SAMPL) tests revealed dose-related increases in sequence alterations in plantlets exposed to
A G Levis et al.
Cancer research, 38(1), 110-116 (1978-01-01)
Treatments for 1 to 4 hr with 10-4 m potassium dichromate, a soluble hexavalent chromium salt with a strong oxidizing power, markedly reduce DNA and RNA accumulation rates in hamster fibroblasts grown in vitro (BHK line), as shown by quantitative
Nanoscale zero-valent iron supported on mesoporous silica: Characterization and reactivity for Cr(VI) removal from aqueous solution.
Petala E, et al.
Journal of Hazardous Materials, 261, 295-306 (2013)
Eduardo Molina-Jijón et al.
Free radical biology & medicine, 51(8), 1543-1557 (2011-08-16)
We report the role of mitochondria in the protective effects of curcumin, a well-known direct and indirect antioxidant, against the renal oxidant damage induced by the hexavalent chromium [Cr(VI)] compound potassium dichromate (K(2)Cr(2)O(7)) in rats. Curcumin was given daily by
An accurate determination of the crystal structure of triclinic potassium dichromate, K2Cr2O7.
Brandon JK and Brown ID.
Canadian Journal of Chemistry, 46(6), 933-941 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service