Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium dichromate
Synonym(s):
Potassium bichromate, Potassium dichromate
Linear Formula:
K
2
Cr
2
O
7
CAS Number:
7778-50-9
Molecular Weight:
294.18
EC Number:
231-906-6
Products
Product Category
salts (5)
Brand
Sigma-Aldrich (4)
Supelco (6)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Reagent Type
Special Grade
ACS reagent (1)
Reference Material (1)
certified reference material (1)
Technique
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
207802
Potassium dichromate,
ACS reagent, ≥99.0%
Expand
View Pricing
P5271
Potassium dichromate,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
1.04864
Potassium dichromate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.02403
Potassium dichromate,
volumetric standard, secondary reference material for redox titration, traceable to NIST SRM Certipur
®
Reag. USP
Expand
View Pricing
P2588
Potassium dichromate,
BioXtra, ≥99.5%
Expand
View Pricing
483044
Potassium dichromate,
99.98% trace metals basis
Expand
View Pricing
1.04865
Potassium dichromate,
for analysis (max. 0.000001% Hg) EMSURE
®
ACS,ISO
Expand
View Pricing
42019
Potassium dichromate,
reference material for titrimetry, certified by BAM, >99.5%
Expand
View Pricing
NIST136F
Potassium dichromate,
NIST
®
SRM
®
136f, oxidimetric standard
Expand
View Pricing
1.93215
Potassium dichromate,
for analysis EMPARTA
®
ACS
Expand
View Pricing
1.93615
Potassium dichromate,
EMPLURA
®
Expand
View Pricing
24-4480
Potassium dichromate,
JIS special grade, ≥99.5%
Expand
View Pricing