Skip to Content
Merck
All Photos(6)

Key Documents

F2877

Sigma-Aldrich

Iron(III) chloride hexahydrate

greener alternative

reagent grade, ≥98%, chunks

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
chunks

grade

reagent grade

Quality Level

Agency

suitable for SM 5210

vapor pressure

1 mmHg ( 194 °C)

Assay

≥98%

form

chunks

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

280-285 °C (lit.)

mp

37 °C (lit.)

greener alternative category

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. The iron catalysts are known to be inexpensive and environmentally benign. Click here for more information.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Enver Fehim Kocpinar et al.
Biological trace element research, 195(1), 135-141 (2019-07-17)
Despite the fact that iron represents a crucial element for the catalysis of many metabolic reactions, its accumulation in the cell leads to the production of reactive oxygen species (ROS), provoking pathological conditions such as cancer, cardiovascular diseases, diabetes, neurodegenerative
Mario Leutert et al.
Molecular systems biology, 15(12), e9021-e9021 (2019-12-31)
Recent developments in proteomics have enabled signaling studies where > 10,000 phosphosites can be routinely identified and quantified. Yet, current analyses are limited in throughput, reproducibility, and robustness, hampering experiments that involve multiple perturbations, such as those needed to map kinase-substrate
Sanjay Kashyap et al.
ACS nano, 8(9), 9097-9106 (2014-08-28)
Biomineralization proteins are widely used as templating agents in biomimetic synthesis of a variety of organic-inorganic nanostructures. However, the role of the protein in controlling the nucleation and growth of biomimetic particles is not well understood, because the mechanism of
Yuchen Guo et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-28)
Pterygoplichthys disjunctivus, locally named the armoured catfish, is a by-catch of tilapia fishing that accounts for up to 80% of total captured fish in the Adolfo Lopez Mateos dam, in Michoacán, México, affecting the economy of its surrounding communities. This
Dattatraya H Dethe et al.
Organic letters, 15(3), 429-431 (2013-01-16)
A novel approach was developed for the synthesis of highly substituted indene derivatives, using an FeCl(3) catalyzed Prins-type cyclization reaction which was further applied in the total synthesis of jungianol and epi-jungianol.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service